Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SGCE SIRNA Assays ›
Gene Symbol:
SGCE
Gene Name:
sarcoglycan epsilon
Gene Aliases:
DYT11, ESG
Chromosome Location:
Chr.7: 94585224 - 94656209 on Build GRCh38
UniGene ID:
Hs.371199
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s17043
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SGCE
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001099400.1
NP_001092870.1
8
1214
NM_001099401.1
NP_001092871.1
9
1241
NM_001301139.1
NP_001288068.1
8
1118
NM_003919.2
NP_003910.1
9
1241
XM_005250675.4
XP_005250732.1
10
1343
XM_005250677.4
XP_005250734.1
9
1297
XM_011516663.2
XP_011514965.1
10
1324
XM_011516664.2
XP_011514966.1
9
1297
XM_011516665.2
XP_011514967.1
9
1219
XM_011516666.2
XP_011514968.1
8
1192
XM_011516667.2
XP_011514969.1
11
1356
XM_011516669.2
XP_011514971.1
10
1262
XM_017012763.1
XP_016868252.1
10
1192
XM_017012764.1
XP_016868253.1
8
1192
XM_017012765.1
XP_016868254.1
11
1355
XM_017012766.1
XP_016868255.1
9
1165
XM_017012767.1
XP_016868256.1
9
1248
XM_017012768.1
XP_016868257.1
10
1262
XM_017012769.1
XP_016868258.1
9
1235
GenBank mRNA
AB117974.1
-
8
1066
AF031920.1
-
3
254
AF036364.1
-
9
1169
AF516515.1
-
9
1130
AJ000534.1
-
9
1198
AK295021.1
-
10
1306
AK298220.1
-
8
1075
AK313438.1
-
9
1180
AY359042.1
-
9
1147
BC021709.2
-
9
1181
4822278
KU178449.1
-
9
1130
KU178450.1
-
8
1103
Back To Top
More Information
Panther Classification:
Molecular Function -
cadherin
Gene Ontology Categories:
Function(s)
muscle system process
cell-matrix adhesion
muscle organ development
membrane organization
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wrkvw:80/100.66.78.186:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0