Search
Search

CCRL2| Gene Symbol: CCRL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130910.1 | NP_001124382.1 | 2 | 232 | ||
| NM_003965.4 | NP_003956.2 | 2 | 368 | |||
| XM_011534208.1 | XP_011532510.1 | 3 | 312 | |||
| XM_011534209.1 | XP_011532511.1 | 3 | 235 | |||
| XM_017007436.1 | XP_016862925.1 | 2 | 228 | |||
| GenBank mRNA | AF015524.1 | - | 2 | 166 | ||
| AF015525.1 | - | 2 | 410 | |||
| AJ344142.1 | - | 1 | 83 | |||
| AK296673.1 | - | 2 | 232 | |||
| AK313312.1 | - | 2 | 156 | |||
| BC071682.1 | - | 2 | 321 | 5182006 | ||
| BC096075.1 | - | 2 | 148 | 40000578 | ||
| BC096076.3 | - | 2 | 148 | 40000580 | ||