Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SYT7 SIRNA Assays ›
          Gene Symbol:
          SYT7
          Gene Name:
          synaptotagmin 7
          Gene Aliases:
          IPCA-7, IPCA7, PCANAP7, SYT-VII, SYTVII
          Chromosome Location:
          Chr.11: 61513716 - 61588404 on Build GRCh38
          UniGene ID:
          Hs.131188
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s17291

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SYT7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001252065.1 NP_001238994.1 8 1130
          NM_001300773.1 NP_001287702.1 8 1037
          NM_004200.3 NP_004191.2 7 905
          XM_005274383.4 XP_005274440.1 11 1792
          XM_005274384.2 XP_005274441.1 10 1796
          XM_005274385.4 XP_005274442.1 10 1562
          XM_005274387.4 XP_005274444.1 5 1829
          XM_005274390.4 XP_005274447.1 4 868
          XM_006718736.3 XP_006718799.1 4 839
          XM_011545335.2 XP_011543637.1 11 1758
          XM_011545336.2 XP_011543638.1 10 1633
          XM_011545337.2 XP_011543639.1 10 1625
          XM_011545338.2 XP_011543640.1 10 1530
          XM_011545339.2 XP_011543641.1 9 1257
          XM_011545340.2 XP_011543642.1 8 1123
          XM_011545341.2 XP_011543643.1 8 1027
          XM_011545343.2 XP_011543645.1 7 891
          GenBank mRNA AF038535.1 - 7 898
          BC125170.1 - 7 863 40121712

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_021659, XM_006231069, XM_006231065, XM_006231066, XM_006231067, and others.
          This siRNA also targets the following transcripts in Mouse: NM_173068, NM_018801, NM_173067, AB026804, BC105660, and others.

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          plasma membrane repair
          vesicle fusion
          phagocytosis
          regulation of calcium ion-dependent exocytosis
          synaptic vesicle recycling
          positive regulation of calcium ion-dependent exocytosis
          regulation of bone remodeling
          calcium ion-regulated exocytosis of neurotransmitter
          regulation of phagocytosis
          regulation of insulin secretion
          regulation of glucagon secretion
          vesicle-mediated cholesterol transport
          phagosome-lysosome fusion
          short-term synaptic potentiation
          calcium ion regulated lysosome exocytosis
          calcium ion binding
          protein binding
          calmodulin binding
          calcium-dependent phospholipid binding
          phosphatidylinositol-4,5-bisphosphate binding
          syntaxin binding
          clathrin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0