Search
Search

Olr844
LOC100912374
Olr848| Gene Symbol: Olr844 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000855.1 | NP_001000855.1 | 1 | 598 | ||
| XM_008763721.1 | XP_008761943.1 | 1 | 682 | |||
| Gene Symbol: LOC100912374 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006238174.2 | XP_006238236.1 | 1 | 682 | ||
| Gene Symbol: Olr848 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000402.1 | NP_001000402.1 | 1 | 598 | ||