Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LRRFIP2 SIRNA Assays ›
Gene Symbol:
LRRFIP2
Gene Name:
LRR binding FLII interacting protein 2
Gene Aliases:
HUFI-2
Chromosome Location:
Chr.3: 37052626 - 37176817 on Build GRCh38
UniGene ID:
Hs.740522
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s17602
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LRRFIP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134369.2
NP_001127841.1
4
585
NM_001282691.1
NP_001269620.1
4
337
NM_006309.3
NP_006300.1
3
284
NM_017724.2
NP_060194.1
4
585
XM_005265539.2
XP_005265596.1
4
1284
XM_005265540.1
XP_005265597.1
5
567
XM_005265550.1
XP_005265607.1
4
552
XM_005265551.1
XP_005265608.1
4
552
XM_005265553.1
XP_005265610.1
4
552
XM_005265554.2
XP_005265611.1
4
1284
XM_005265556.1
XP_005265613.1
4
552
XM_006713385.1
XP_006713448.1
4
552
XM_006713386.1
XP_006713449.1
4
552
XM_006713387.1
XP_006713450.1
4
552
XM_006713388.1
XP_006713451.1
4
552
XM_006713389.1
XP_006713452.1
4
552
XM_006713390.1
XP_006713453.1
5
595
XM_006713392.1
XP_006713455.1
4
552
XM_006713393.1
XP_006713456.1
4
552
XM_006713394.1
XP_006713457.1
4
552
XM_006713395.1
XP_006713458.1
4
552
XM_006713396.1
XP_006713459.1
4
552
XM_006713397.1
XP_006713460.1
4
552
XM_006713400.1
XP_006713463.1
4
552
XM_006713404.1
XP_006713467.1
4
552
XM_011534217.1
XP_011532519.1
4
552
XM_011534218.1
XP_011532520.1
5
595
XM_011534219.1
XP_011532521.1
5
663
XM_011534221.1
XP_011532523.1
4
552
XM_011534222.1
XP_011532524.1
4
552
XM_011534229.1
XP_011532531.1
5
595
XM_017007465.1
XP_016862954.1
4
552
XM_017007466.1
XP_016862955.1
4
552
XM_017007467.1
XP_016862956.1
4
552
XM_017007468.1
XP_016862957.1
4
552
XM_017007469.1
XP_016862958.1
4
552
XM_017007470.1
XP_016862959.1
4
552
XM_017007471.1
XP_016862960.1
4
552
XM_017007472.1
XP_016862961.1
4
552
XM_017007473.1
XP_016862962.1
4
552
XM_017007474.1
XP_016862963.1
4
552
XM_017007475.1
XP_016862964.1
4
1284
XM_017007476.1
XP_016862965.1
5
595
XM_017007477.1
XP_016862966.1
4
1284
XM_017007478.1
XP_016862967.1
4
552
XM_017007479.1
XP_016862968.1
4
552
XM_017007480.1
XP_016862969.1
4
796
XM_017007481.1
XP_016862970.1
4
552
XM_017007482.1
XP_016862971.1
4
552
XM_017007483.1
XP_016862972.1
4
1284
XM_017007484.1
XP_016862973.1
5
595
GenBank mRNA
AF115509.1
-
3
243
AK000255.1
-
3
270
AK291514.1
-
4
562
AK302282.1
-
4
311
BC053668.1
-
4
542
6146978
DB033703.1
-
4
338
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
biological_process
Wnt signaling pathway
protein binding
LRR domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0