Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LRRFIP2 SIRNA Assays ›
Gene Symbol:
LRRFIP2
Gene Name:
LRR binding FLII interacting protein 2
Gene Aliases:
HUFI-2
Chromosome Location:
Chr.3: 37052626 - 37176817 on Build GRCh38
UniGene ID:
Hs.740522
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s17604
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LRRFIP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134369.2
NP_001127841.1
14
1511
NM_001282691.1
NP_001269620.1
18
1500
NM_006309.3
NP_006300.1
27
2101
NM_017724.2
NP_060194.1
13
1439
XM_005265539.2
XP_005265596.1
28
3101
XM_005265540.1
XP_005265597.1
29
2384
XM_005265550.1
XP_005265607.1
20
1937
XM_005265551.1
XP_005265608.1
19
1826
XM_005265553.1
XP_005265610.1
16
1673
XM_005265554.2
XP_005265611.1
16
2405
XM_005265556.1
XP_005265613.1
15
1601
XM_006713385.1
XP_006713448.1
28
2366
XM_006713386.1
XP_006713449.1
27
2327
XM_006713387.1
XP_006713450.1
27
2324
XM_006713388.1
XP_006713451.1
27
2321
XM_006713389.1
XP_006713452.1
27
2318
XM_006713390.1
XP_006713453.1
28
2361
XM_006713392.1
XP_006713455.1
27
2297
XM_006713393.1
XP_006713456.1
27
2279
XM_006713394.1
XP_006713457.1
26
2276
XM_006713395.1
XP_006713458.1
27
2276
XM_006713396.1
XP_006713459.1
26
2261
XM_006713397.1
XP_006713460.1
25
2153
XM_006713400.1
XP_006713463.1
17
1718
XM_006713404.1
XP_006713467.1
14
1451
XM_011534217.1
XP_011532519.1
28
2369
XM_011534218.1
XP_011532520.1
29
2412
XM_011534219.1
XP_011532521.1
29
2480
XM_011534221.1
XP_011532523.1
26
2261
XM_011534222.1
XP_011532524.1
26
2174
XM_011534229.1
XP_011532531.1
18
1761
XM_017007465.1
XP_016862954.1
25
2216
XM_017007466.1
XP_016862955.1
26
2210
XM_017007467.1
XP_016862956.1
26
2204
XM_017007468.1
XP_016862957.1
23
2108
XM_017007469.1
XP_016862958.1
25
2102
XM_017007470.1
XP_016862959.1
23
2066
XM_017007471.1
XP_016862960.1
18
1775
XM_017007472.1
XP_016862961.1
18
1733
XM_017007473.1
XP_016862962.1
17
1703
XM_017007474.1
XP_016862963.1
17
1682
XM_017007475.1
XP_016862964.1
17
2414
XM_017007476.1
XP_016862965.1
17
1716
XM_017007477.1
XP_016862966.1
16
2342
XM_017007478.1
XP_016862967.1
15
1580
XM_017007479.1
XP_016862968.1
16
1580
XM_017007480.1
XP_016862969.1
15
1824
XM_017007481.1
XP_016862970.1
15
1571
XM_017007482.1
XP_016862971.1
14
1508
XM_017007483.1
XP_016862972.1
14
2210
XM_017007484.1
XP_016862973.1
15
1521
GenBank mRNA
AF115509.1
-
27
2060
AK000255.1
-
12
1124
AK024692.1
-
6
457
AK124432.1
-
5
3235
AK291514.1
-
14
1488
AK302282.1
-
18
1474
BC053668.1
-
13
1396
6146978
BX648794.1
-
10
890
CR749705.1
-
4
1828
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
biological_process
Wnt signaling pathway
protein binding
LRR domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0