Search
Search

Hmgb2
Hmgb2l1
RGD1559962| Gene Symbol: Hmgb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017187.2 | NP_058883.1 | 4 | 482 | ||
| GenBank mRNA | BC078866.1 | - | 3 | 340 | 7114119 | |
| BC089854.1 | - | 4 | 482 | 7380208 | ||
| BC107455.1 | - | 4 | 511 | 7377505 | ||
| D84418.1 | - | 4 | 404 | |||
| FQ225084.1 | - | 4 | 446 | |||
| FQ229618.1 | - | 4 | 419 | |||
| FQ230440.1 | - | 4 | 446 | |||
| FQ233747.1 | - | 4 | 420 | |||
| Gene Symbol: Hmgb2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_573272.5 | XP_573272.3 | 1 | 419 | ||
| Gene Symbol: RGD1559962 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758645.1 | XP_008756867.1 | 1 | 1834 | ||