Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRKRIP1 SIRNA Assays ›
Gene Symbol:
Prkrip1
Gene Name:
Prkr interacting protein 1 (IL11 inducible)
Gene Aliases:
-
Chromosome Location:
Chr.12: 23514987 - 23533278 on Build Rnor_6.0
UniGene ID:
Rn.44767
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s176548
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Prkrip1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001098793.1
NP_001092263.1
2
226
GenBank mRNA
BC158549.1
-
2
226
8361725
DY314400.1
-
2
226
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Human: NM_024653, BC014298, AF178686, AK294664, BX459931, and others.
This siRNA also targets the following transcripts in Mouse: NM_025774, AK018438, BC027503, AK010961, AK010359, and others.
Gene Ontology Categories:
Function(s)
Process(es)
renal system process
negative regulation of protein kinase activity
negative regulation of phosphorylation
double-stranded RNA binding
protein kinase inhibitor activity
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline