Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PKD1P6-NPIPP1 SIRNA Assays ›
          Gene Symbol:
          PKD1P6-NPIPP1 LOC105376752 NPIPA1 NPIPA2 NPIPA3 NPIPA5 NPIPA7 NPIPA8 PKD1P1
          Gene Name:
          PKD1P6-NPIPP1 readthrough
          nuclear pore complex-interacting protein family member A5
          nuclear pore complex interacting protein family member A1
          nuclear pore complex interacting protein family member A2
          nuclear pore complex interacting protein family member A3
          nuclear pore complex interacting protein family member A5
          nuclear pore complex interacting protein family member A7
          nuclear pore complex interacting protein family member A8
          polycystin 1, transient receptor potential channel interacting pseudogene 1
          Gene Aliases:
          -
          -
          NPIP, NPIPA, morpheus
          NPIP
          -
          NPIP
          -
          LCR16a9
          HG1, NPIP
          Chromosome Location:
          Chr.16: 15104723 - 15131601 on Build GRCh38
          Chromosome Location:
          Chr.16: 18359533 - 18372055 on Build GRCh38
          Chromosome Location:
          Chr.16: 14935258 - 14952074 on Build GRCh38
          Chromosome Location:
          Chr.16: 14745595 - 14765448 on Build GRCh38
          Chromosome Location:
          Chr.16: 14706473 - 14726338 on Build GRCh38
          Chromosome Location:
          Chr.16: 15363624 - 15381478 on Build GRCh38
          Chromosome Location:
          Chr.16: 16376940 - 16393972 on Build GRCh38
          Chromosome Location:
          Chr.16: 18317919 - 18334967 on Build GRCh38
          Chromosome Location:
          Chr.16: 16317609 - 16350608 on Build GRCh38
          UniGene ID:
          Hs.634612
          -
          Hs.626677
          -
          -
          Hs.676266
          Hs.634612
          Hs.636569
          Hs.636569
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s17752

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PKD1P6-NPIPP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_123721.1 25 4689
          NR_123722.1 15 2550
          GenBank mRNA AK131084.1 - 20 5053
          AK131085.1 - 19 4959
          AK302708.1 - 15 2550
          Gene Symbol: LOC105376752
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_011522797.2 XP_011521099.1 5 619
          Gene Symbol: NPIPA1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_006985.2 NP_008916.2 6 596
          XM_011522748.2 XP_011521050.1 6 693
          XM_011522749.2 XP_011521051.1 7 805
          XM_011522750.2 XP_011521052.1 5 611
          XM_011522751.1 XP_011521053.1 6 692
          XM_017023891.1 XP_016879380.1 7 805
          GenBank mRNA AF132984.1 - 6 596
          AK160377.1 - 25 4783
          AK294177.1 - 2 73
          KF175165.1 - 6 632
          KF175166.1 - 5 524
          KF175167.1 - 6 632
          KF175169.1 - 6 596
          KF175208.1 - 6 743
          KF175209.1 - 7 814
          KF175225.1 - 6 596
          Gene Symbol: NPIPA2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277324.1 NP_001264253.1 6 596
          XM_005255489.3 XP_005255546.1 8 3015
          XM_005255492.4 XP_005255549.1 6 1000
          XM_006720917.3 XP_006720980.2 8 3059
          XM_011522587.1 XP_011520889.1 8 3059
          XM_011522593.1 XP_011520895.1 8 3059
          XM_011522594.2 XP_011520896.1 8 3059
          XM_011522595.1 XP_011520897.1 8 3059
          XM_011522596.1 XP_011520898.1 5 515
          XM_011522597.1 XP_011520899.1 7 2951
          GenBank mRNA KF175213.1 - 5 289
          Gene Symbol: NPIPA3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277323.1 NP_001264252.1 6 596
          XM_005255484.3 XP_005255541.1 7 684
          XM_005255485.4 XP_005255542.1 6 1000
          XM_011522576.2 XP_011520878.2 8 3049
          XM_011522577.2 XP_011520879.2 8 2970
          XM_011522578.2 XP_011520880.2 6 692
          XM_011522580.2 XP_011520882.2 6 623
          XM_011522581.1 XP_011520883.1 8 3059
          XM_011522583.2 XP_011520885.1 8 3059
          XM_011522584.1 XP_011520886.1 8 3059
          XM_011522585.1 XP_011520887.1 5 515
          XM_011522586.1 XP_011520888.1 7 2951
          XM_017023553.1 XP_016879042.1 8 3059
          XM_017023554.1 XP_016879043.1 7 2972
          Gene Symbol: NPIPA5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277325.1 NP_001264254.1 6 596
          XM_011522329.2 XP_011520631.1 8 1007
          XM_011522336.2 XP_011520638.1 8 1000
          XM_011522337.1 XP_011520639.1 5 515
          GenBank mRNA CB123100.1 - 3 274
          EL734608.1 - 6 596
          KF175168.1 - 6 593
          KF175170.1 - 6 596
          KF175171.1 - 6 590
          KF175172.1 - 6 596
          KF175173.1 - 4 371
          Gene Symbol: NPIPA7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282507.1 NP_001269436.1 6 596
          XM_005255735.4 XP_005255792.1 6 718
          XM_011522339.2 XP_011520641.1 6 644
          XM_017022833.1 XP_016878322.1 7 669
          XM_017022834.1 XP_016878323.1 7 721
          XM_017022835.1 XP_016878324.1 7 782
          XM_017022836.1 XP_016878325.1 6 589
          XM_017022837.1 XP_016878326.1 7 669
          XM_017022838.1 XP_016878327.1 6 1064
          XM_017022839.1 XP_016878328.1 6 561
          XM_017022840.1 XP_016878329.1 6 609
          GenBank mRNA BI046418.1 - 3 247
          Gene Symbol: NPIPA8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282511.1 NP_001269440.1 6 596
          XM_005255738.4 XP_005255795.1 6 718
          XM_006720830.3 XP_006720893.1 6 1616
          XM_011522342.2 XP_011520644.1 6 645
          XM_017022841.1 XP_016878330.1 7 669
          XM_017022842.1 XP_016878331.1 7 721
          XM_017022843.1 XP_016878332.1 7 782
          XM_017022844.1 XP_016878333.1 6 589
          XM_017022845.1 XP_016878334.1 7 669
          XM_017022846.1 XP_016878335.1 6 561
          XM_017022847.1 XP_016878336.1 6 609
          GenBank mRNA BY996642.1 - 2 177
          Gene Symbol: PKD1P1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK298381.1 - 4 354
          BC039707.1 - 7 915 4479933
          BC046145.1 - 8 1007
          KF175174.1 - 6 632
          KF175210.1 - 7 878
          KF175211.1 - 8 952
          KF175212.1 - 9 765
          KF175214.1 - 7 814
          KF175219.1 - 6 782

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          protein transport
          mRNA transport
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0