Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SRSF11 SIRNA Assays ›
          Gene Symbol:
          SRSF11 PLEKHA5 SRSF11P1
          Gene Name:
          serine and arginine rich splicing factor 11
          pleckstrin homology domain containing A5
          serine and arginine rich splicing factor 11 pseudogene 1
          Gene Aliases:
          NET2, SFRS11, dJ677H15.2, p54
          PEPP-2, PEPP2
          -
          Chromosome Location:
          Chr.1: 70205682 - 70252020 on Build GRCh38
          Chromosome Location:
          Chr.12: 19129680 - 19376400 on Build GRCh38
          Chromosome Location:
          Chr.12: 19306671 - 19308236 on Build GRCh38
          UniGene ID:
          Hs.479693
          Hs.188614
          Hs.655546
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID s17771

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SRSF11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001190987.1 NP_001177916.1 7 765
          NM_004768.3 NP_004759.1 7 768
          XM_005271338.3 XP_005271395.1 7 759
          XM_005271339.3 XP_005271396.1 7 761
          XM_011542427.2 XP_011540729.1 6 921
          XM_011542428.2 XP_011540730.1 6 4157
          XM_011542431.2 XP_011540733.1 7 1921
          XM_017002811.1 XP_016858300.1 7 759
          XM_017002812.1 XP_016858301.1 7 759
          XM_017002813.1 XP_016858302.1 7 761
          XM_017002814.1 XP_016858303.1 5 1890
          XM_017002815.1 XP_016858304.1 7 3502
          XM_017002816.1 XP_016858305.1 8 3636
          XM_017002817.1 XP_016858306.1 8 3210
          XM_017002818.1 XP_016858307.1 7 1038
          XM_017002819.1 XP_016858308.1 8 1147
          XM_017002820.1 XP_016858309.1 7 3865
          XM_017002821.1 XP_016858310.1 7 1038
          XM_017002822.1 XP_016858311.1 5 2253
          XM_017002823.1 XP_016858312.1 6 1598
          XM_017002824.1 XP_016858313.1 8 3636
          XM_017002825.1 XP_016858314.1 8 3210
          XM_017002826.1 XP_016858315.1 7 1038
          XM_017002827.1 XP_016858316.1 8 1147
          XM_017002828.1 XP_016858317.1 9 2981
          XM_017002829.1 XP_016858318.1 9 3017
          XM_017002830.1 XP_016858319.1 7 4142
          XM_017002831.1 XP_016858320.1 8 3210
          XM_017002832.1 XP_016858321.1 7 1038
          XM_017002833.1 XP_016858322.1 8 1147
          XM_017002834.1 XP_016858323.1 6 1880
          GenBank mRNA AK301670.1 - 7 936
          AL832279.1 - 4 1027
          BC002784.2 - 6 779
          BC009840.2 - 6 762
          BC017359.2 - 6 885
          BC022815.1 - 7 765
          BC032727.1 - 7 752
          BC040436.1 - 7 723 5518580
          BX640645.1 - 5 2005
          BX641004.1 - 3 188
          M74002.1 - 7 727
          Gene Symbol: PLEKHA5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC000969.2 - 8 2143
          Gene Symbol: SRSF11P1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK021937.1 - 1 392

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          termination of RNA polymerase II transcription
          mRNA processing
          RNA export from nucleus
          mRNA export from nucleus
          RNA splicing
          mRNA 3'-end processing
          nucleotide binding
          protein binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0