Search
Search

Hyal5| Gene Symbol: Hyal5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024321.1 | NP_001019492.1 | 4 | 1495 | ||
| XM_006236166.2 | XP_006236228.1 | 5 | 1613 | |||
| XM_008762806.1 | XP_008761028.1 | 6 | 1841 | |||
| XM_008762807.1 | XP_008761029.1 | 5 | 1748 | |||
| XM_008762808.1 | XP_008761030.1 | 5 | 1663 | |||
| GenBank mRNA | BC079447.1 | - | 6 | 1818 | ||
| BC091219.1 | - | 4 | 1495 | 7112082 | ||