Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NRXN1 SIRNA Assays ›
Gene Symbol:
NRXN1
Gene Name:
neurexin 1
Gene Aliases:
Hs.22998, PTHSL2, SCZD17
Chromosome Location:
Chr.2: 49918503 - 51032536 on Build GRCh38
UniGene ID:
Hs.637685
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s17947
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NRXN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135659.1
NP_001129131.1
19
4976
NM_004801.4
NP_004792.1
18
4856
NM_138735.2
NP_620072.1
2
1081
XM_005264642.3
XP_005264699.1
18
4877
XM_005264643.3
XP_005264700.1
17
4832
XM_006712137.3
XP_006712200.1
18
4877
XM_006712140.3
XP_006712203.2
19
4904
XM_011533167.2
XP_011531469.1
18
4877
XM_011533171.2
XP_011531473.1
18
4856
XM_011533172.2
XP_011531474.1
17
4850
XM_011533174.2
XP_011531476.1
17
4832
XM_011533175.2
XP_011531477.1
16
4820
XM_011533177.2
XP_011531479.1
18
4877
XM_011533178.2
XP_011531480.1
15
4787
XM_011533180.2
XP_011531482.1
18
4877
XM_011533183.1
XP_011531485.1
12
2559
XM_017005303.1
XP_016860792.1
19
4904
XM_017005304.1
XP_016860793.1
19
4901
XM_017005305.1
XP_016860794.1
19
4904
XM_017005306.1
XP_016860795.1
18
4892
XM_017005307.1
XP_016860796.1
18
4886
XM_017005308.1
XP_016860797.1
19
4883
XM_017005309.1
XP_016860798.1
18
4877
XM_017005310.1
XP_016860799.1
17
4874
XM_017005311.1
XP_016860800.1
18
4859
XM_017005312.1
XP_016860801.1
18
4853
XM_017005313.1
XP_016860802.1
18
4856
XM_017005314.1
XP_016860803.1
16
4844
XM_017005315.1
XP_016860804.1
17
4850
XM_017005316.1
XP_016860805.1
17
4841
XM_017005317.1
XP_016860806.1
17
4844
XM_017005318.1
XP_016860807.1
17
4832
XM_017005319.1
XP_016860808.1
17
4829
XM_017005320.1
XP_016860809.1
17
4829
XM_017005321.1
XP_016860810.1
19
4904
XM_017005322.1
XP_016860811.1
19
4904
XM_017005323.1
XP_016860812.1
14
4790
XM_017005324.1
XP_016860813.1
17
4850
XM_017005325.1
XP_016860814.1
17
4850
XM_017005326.1
XP_016860815.1
17
4838
XM_017005327.1
XP_016860816.1
17
4832
XM_017005328.1
XP_016860817.1
16
4805
XM_017005329.1
XP_016860818.1
19
4904
XM_017005330.1
XP_016860819.1
14
4790
XM_017005331.1
XP_016860820.1
14
4775
XM_017005332.1
XP_016860821.1
13
4745
XM_017005333.1
XP_016860822.1
13
4745
XM_017005334.1
XP_016860823.1
15
2725
XM_017005335.1
XP_016860824.1
2
6071
XM_017005336.1
XP_016860825.1
2
6072
XM_017005337.1
XP_016860826.1
2
6071
GenBank mRNA
AB011150.2
-
16
3538
AB035356.1
-
17
3571
AF064842.1
-
2
1080
AK093260.1
-
12
2559
BC125179.1
-
3
137
BC125180.1
-
3
500
BC150247.1
-
16
3559
9007472
EF539882.1
-
19
4839
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neuron cell-cell adhesion
chemical synaptic transmission
neurotransmitter secretion
axon guidance
synapse assembly
learning
adult behavior
social behavior
vocal learning
neuromuscular process controlling balance
positive regulation of synapse assembly
positive regulation of synaptic transmission, glutamatergic
prepulse inhibition
regulation of insulin secretion involved in cellular response to glucose stimulus
vocalization behavior
positive regulation of synapse maturation
postsynaptic membrane assembly
gephyrin clustering involved in postsynaptic density assembly
neuroligin clustering involved in postsynaptic membrane assembly
postsynaptic density protein 95 clustering
positive regulation of excitatory postsynaptic potential
regulation of grooming behavior
receptor activity
calcium channel regulator activity
calcium ion binding
protein binding
acetylcholine receptor binding
cell adhesion molecule binding
neuroligin family protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline