Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KDM1A SIRNA Assays ›
Gene Symbol:
Kdm1a
Gene Name:
lysine (K)-specific demethylase 1A
Gene Aliases:
Aof2, Kdm1, RGD1562975
Chromosome Location:
Chr.5: 154909003 - 154965171 on Build Rnor_6.0
UniGene ID:
Rn.203461
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s179475
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Kdm1a
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130098.1
NP_001123570.1
14, 15
1874
GenBank mRNA
EF688598.1
-
14, 15
1874
Back To Top
More Information
Panther Classification:
Molecular Function -
oxidase
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
alternative mRNA splicing, via spliceosome
in utero embryonic development
regulation of transcription from RNA polymerase II promoter
protein demethylation
cell proliferation
regulation of primitive erythrocyte differentiation
positive regulation of neuron projection development
response to organic cyclic compound
pituitary gland development
cerebral cortex development
granulocyte differentiation
negative regulation of protein binding
histone H3-K9 demethylation
histone H3-K4 demethylation
positive regulation of chromatin binding
neuron maturation
negative regulation of sequence-specific DNA binding transcription factor activity
negative regulation of DNA damage response, signal transduction by p53 class mediator
positive regulation of erythrocyte differentiation
positive regulation of megakaryocyte differentiation
positive regulation of cell size
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
guanine metabolic process
positive regulation of hormone biosynthetic process
positive regulation of sequence-specific DNA binding transcription factor activity
negative regulation of histone H3-K4 methylation
negative regulation of histone H3-K9 methylation
muscle cell development
oxidation-reduction process
response to fungicide
cellular response to cAMP
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
positive regulation of neural precursor cell proliferation
positive regulation of stem cell proliferation
RNA polymerase II transcription factor binding
p53 binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
oxidoreductase activity
enzyme binding
ligand-dependent nuclear receptor transcription coactivator activity
demethylase activity
histone demethylase activity
histone demethylase activity (H3-K4 specific)
histone demethylase activity (H3-K9 specific)
histone demethylase activity (H3-dimethyl-K4 specific)
MRF binding
transcription regulatory region DNA binding
flavin adenine dinucleotide binding
androgen receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-fhf6r:80/100.66.78.180:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0