Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HAND2 SIRNA Assays ›
          Gene Symbol:
          HAND2
          Gene Name:
          heart and neural crest derivatives expressed 2
          Gene Aliases:
          DHAND2, Hed, Thing2, bHLHa26, dHand
          Chromosome Location:
          Chr.4: 173526501 - 173530227 on Build GRCh38
          UniGene ID:
          Hs.388245
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s18133

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HAND2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_021973.2 NP_068808.1 1, 2 1502
          GenBank mRNA AF087940.1 - 1, 2 468
          AK122739.1 - 2, 3 897
          BC101406.1 - 2, 3 543 40003700
          BC101974.1 - 2, 3 551
          BC109383.1 - 2, 3 316
          BF313018.1 - 1, 2 188
          FJ226608.1 - 1, 2 1504

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          in utero embryonic development
          heart looping
          suckling behavior
          cardiac right ventricle formation
          cardiac neural crest cell migration involved in outflow tract morphogenesis
          regulation of secondary heart field cardioblast proliferation
          apoptotic process involved in heart morphogenesis
          noradrenergic neuron differentiation
          transcription from RNA polymerase II promoter
          heart development
          adult heart development
          mesenchymal cell proliferation
          positive regulation of cardiac muscle hypertrophy
          positive regulation of gene expression
          negative regulation of gene expression
          negative regulation of cardiac muscle cell apoptotic process
          regulation of tissue remodeling
          odontogenesis of dentin-containing tooth
          embryonic digit morphogenesis
          negative regulation of DNA binding
          negative regulation of sequence-specific DNA binding transcription factor activity
          tongue development
          negative regulation of osteoblast differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          sympathetic nervous system development
          thymus development
          peripheral nervous system neuron development
          palate development
          cartilage morphogenesis
          coronary artery morphogenesis
          visceral serous pericardium development
          cardiac neural crest cell development involved in outflow tract morphogenesis
          cell proliferation involved in outflow tract morphogenesis
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of p38MAPK cascade
          positive regulation of transcription regulatory region DNA binding
          positive regulation of transcription from RNA polymerase II promoter involved in norepinephrine biosynthetic process
          positive regulation of semaphorin-plexin signaling pathway involved in outflow tract morphogenesis
          RNA polymerase II regulatory region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          AT DNA binding
          transcription coactivator activity
          protein binding
          transcription factor binding
          activating transcription factor binding
          protein homodimerization activity
          transcription regulatory region DNA binding
          protein heterodimerization activity
          E-box binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0