Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ROCK2 SIRNA Assays ›
          Gene Symbol:
          ROCK2
          Gene Name:
          Rho associated coiled-coil containing protein kinase 2
          Gene Aliases:
          ROCK-II
          Chromosome Location:
          Chr.2: 11179759 - 11345407 on Build GRCh38
          UniGene ID:
          Hs.681743
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s18162
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ROCK2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001321643.1 NP_001308572.1 13, 14 1460
          NM_004850.4 NP_004841.2 13, 14 1916
          XM_005246190.3 XP_005246247.1 13, 14 1950
          XM_011510417.2 XP_011508719.1 13, 14 1415
          XM_017005378.1 XP_016860867.1 13, 14 1947
          XM_017005379.1 XP_016860868.1 13, 14 1345
          GenBank mRNA AB014519.1 - 13, 14 1921
          AK310358.1 - 9, 10 1211
          BG539531.1 - 284
          D87931.1 - 13, 14 1467

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00153074_m1

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          cytokinesis
          positive regulation of protein phosphorylation
          protein phosphorylation
          smooth muscle contraction
          I-kappaB kinase/NF-kappaB signaling
          Rho protein signal transduction
          positive regulation of endothelial cell migration
          positive regulation of gene expression
          positive regulation of centrosome duplication
          negative regulation of angiogenesis
          regulation of cell adhesion
          cortical actin cytoskeleton organization
          regulation of actin cytoskeleton organization
          negative regulation of myosin-light-chain-phosphatase activity
          regulation of circadian rhythm
          regulation of keratinocyte differentiation
          vascular endothelial growth factor receptor signaling pathway
          ephrin receptor signaling pathway
          rhythmic process
          centrosome duplication
          regulation of stress fiber assembly
          regulation of focal adhesion assembly
          cellular response to testosterone stimulus
          establishment of protein localization to plasma membrane
          regulation of establishment of endothelial barrier
          negative regulation of bicellular tight junction assembly
          regulation of establishment of cell polarity
          regulation of cell motility
          protein serine/threonine kinase activity
          structural molecule activity
          protein binding
          ATP binding
          Rho GTPase binding
          poly(A) RNA binding
          metal ion binding
          Rho-dependent protein serine/threonine kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0