Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CYCS SIRNA Assays ›
          Gene Symbol:
          Cycs LOC100363502
          Gene Name:
          cytochrome c, somatic
          cytochrome c, somatic-like
          Gene Aliases:
          CYCSA
          -
          Chromosome Location:
          Chr.4: 80331226 - 80333326 on Build Rnor_6.0
          Chromosome Location:
          Chr.18: 62475572 - 62476771 on Build Rnor_6.0
          UniGene ID:
          Rn.161841
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s182848

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cycs
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012839.2 NP_036971.1 3 284
          GenBank mRNA BC081849.1 - 3 284 7192255
          BC089051.1 - 3 295 7305562
          FQ211631.1 - 3 276
          FQ212556.1 - 3 278
          FQ212860.1 - 3 336
          FQ213464.1 - 3 276
          FQ214646.1 - 3 307
          FQ214733.1 - 3 276
          FQ214738.1 - 3 274
          FQ214838.1 - 3 275
          FQ214855.1 - 3 307
          FQ214908.1 - 3 276
          FQ215052.1 - 3 276
          FQ215109.1 - 3 276
          FQ215466.1 - 3 276
          FQ216163.1 - 3 275
          FQ216217.1 - 3 274
          FQ216281.1 - 3 334
          FQ216426.1 - 3 276
          FQ216512.1 - 3 276
          FQ216560.1 - 3 276
          FQ216674.1 - 3 275
          FQ216679.1 - 3 276
          FQ217283.1 - 3 276
          FQ217634.1 - 3 276
          FQ218428.1 - 3 276
          FQ218668.1 - 3 277
          FQ220222.1 - 3 307
          FQ220855.1 - 3 319
          FQ223761.1 - 3 276
          FQ224710.1 - 3 276
          FQ224919.1 - 3 276
          M20622.1 - 2 209
          Gene Symbol: LOC100363502
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_003751799.3 XP_003751847.1 1 280

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          response to ischemia
          mitochondrial electron transport, ubiquinol to cytochrome c
          mitochondrial electron transport, cytochrome c to oxygen
          apoptotic DNA fragmentation
          apoptotic process
          response to oxidative stress
          activation of cysteine-type endopeptidase activity involved in apoptotic process by cytochrome c
          negative regulation of hydrogen peroxide biosynthetic process
          glial cell apoptotic process
          hydrogen peroxide metabolic process
          positive regulation of cellular respiration
          iron ion binding
          lipid binding
          electron carrier activity
          heme binding
          electron transporter, transferring electrons from CoQH2-cytochrome c reductase complex and cytochrome c oxidase complex activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0