Search
Search

Tmem230| Gene Symbol: Tmem230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048043.1 | NP_001041508.1 | 2, 3 | 317 | ||
| XM_006235070.2 | XP_006235132.1 | 3, 4 | 415 | |||
| XM_008762199.1 | XP_008760421.1 | 1, 2 | 116 | |||
| XM_008762200.1 | XP_008760422.1 | 1, 2 | 116 | |||
| GenBank mRNA | BC105614.1 | - | 2, 3 | 317 | 7384127 | |