Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNFAIP3 SIRNA Assays ›
Gene Symbol:
Tnfaip3
Gene Name:
tumor necrosis factor, alpha-induced protein 3
Gene Aliases:
-
Chromosome Location:
Chr.1: 14401103 - 14412822 on Build Rnor_6.0
UniGene ID:
-
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s185976
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tnfaip3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_003748656.3
XP_003748704.2
1
380
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001166402, NM_009397, AK153238, CT010397, AK151921, and others.
Panther Classification:
Molecular Function -
cysteine protease
Gene Ontology Categories:
Function(s)
Process(es)
B-1 B cell homeostasis
response to molecule of bacterial origin
regulation of germinal center formation
negative regulation of chronic inflammatory response
protein deubiquitination
negative regulation of protein ubiquitination
negative regulation of NF-kappaB transcription factor activity
response to muramyl dipeptide
negative regulation of interleukin-1 beta production
negative regulation of interleukin-2 production
negative regulation of interleukin-6 production
negative regulation of tumor necrosis factor production
negative regulation of toll-like receptor 3 signaling pathway
negative regulation of toll-like receptor 5 signaling pathway
protein K11-linked deubiquitination
negative regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of protein catabolic process
negative regulation of cyclin-dependent protein serine/threonine kinase activity
negative regulation of innate immune response
negative regulation of smooth muscle cell proliferation
negative regulation of inflammatory response
negative regulation of B cell activation
negative regulation of cell death
cellular response to hydrogen peroxide
negative regulation of nucleotide-binding oligomerization domain containing 1 signaling pathway
negative regulation of nucleotide-binding oligomerization domain containing 2 signaling pathway
protein K63-linked deubiquitination
protein K48-linked ubiquitination
protein K48-linked deubiquitination
cellular response to lipopolysaccharide
tolerance induction to lipopolysaccharide
establishment of protein localization to vacuole
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
positive regulation of cellular protein catabolic process
positive regulation of hepatocyte proliferation
negative regulation of CD40 signaling pathway
negative regulation of endothelial cell apoptotic process
protease binding
DNA binding
ubiquitin-protein transferase activity
thiol-dependent ubiquitin-specific protease activity
zinc ion binding
kinase binding
ubiquitin binding
protein self-association
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0