Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPP6R2 SIRNA Assays ›
Gene Symbol:
PPP6R2
Gene Name:
protein phosphatase 6 regulatory subunit 2
Gene Aliases:
KIAA0685, PP6R2, SAP190, SAPS2
Chromosome Location:
Chr.22: 50343317 - 50445090 on Build GRCh38
UniGene ID:
Hs.449098
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s18689
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPP6R2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242898.1
NP_001229827.1
4
776
NM_001242899.1
NP_001229828.1
4
776
NM_001242900.1
NP_001229829.1
4
776
NM_014678.4
NP_055493.2
4
776
XM_006724431.2
XP_006724494.1
5
887
XM_006724434.1
XP_006724497.1
6
1036
XM_006724435.2
XP_006724498.1
4
761
XM_011530720.2
XP_011529022.1
3
461
XM_011530721.2
XP_011529023.1
3
461
XM_011530722.2
XP_011529024.1
3
461
XM_011530723.2
XP_011529025.1
3
461
XM_011530724.2
XP_011529026.1
3
461
XM_011530726.2
XP_011529028.1
3
461
XM_011530727.2
XP_011529029.1
3
461
XM_011530728.2
XP_011529030.1
3
461
XM_011530729.2
XP_011529031.1
3
461
XM_011530730.2
XP_011529032.1
3
461
XM_011530731.1
XP_011529033.1
3
461
XM_011530732.2
XP_011529034.1
6
937
XM_011530734.2
XP_011529036.1
5
811
XM_011530735.2
XP_011529037.1
4
761
XM_011530736.2
XP_011529038.1
5
887
XM_011530737.2
XP_011529039.1
6
1036
XM_011530738.2
XP_011529040.1
5
839
XM_011530739.2
XP_011529041.1
3
501
XM_011530740.2
XP_011529042.1
5
909
XM_017029116.1
XP_016884605.1
3
461
XM_017029117.1
XP_016884606.1
7
1086
XM_017029118.1
XP_016884607.1
7
1038
XM_017029119.1
XP_016884608.1
6
960
XM_017029120.1
XP_016884609.1
6
937
XM_017029121.1
XP_016884610.1
4
761
XM_017029122.1
XP_016884611.1
5
839
XM_017029123.1
XP_016884612.1
4
761
XM_017029124.1
XP_016884613.1
5
887
XM_017029125.1
XP_016884614.1
4
761
XM_017029126.1
XP_016884615.1
4
761
XM_017029127.1
XP_016884616.1
5
839
XM_017029128.1
XP_016884617.1
5
887
XM_017029129.1
XP_016884618.1
4
761
XM_017029130.1
XP_016884619.1
4
761
XM_017029131.1
XP_016884620.1
5
887
XM_017029132.1
XP_016884621.1
5
839
XM_017029133.1
XP_016884622.1
4
715
XM_017029134.1
XP_016884623.1
4
715
XM_017029135.1
XP_016884624.1
5
765
XM_017029136.1
XP_016884625.1
4
715
XM_017029137.1
XP_016884626.1
4
715
XM_017029138.1
XP_016884627.1
4
715
GenBank mRNA
AB014585.1
-
4
748
AK302472.1
-
4
687
BC000976.4
-
5
765
2900437
BC006568.2
-
4
762
3532211
BC032664.1
-
4
773
5553142
BC041698.1
-
4
743
5732101
BC052995.2
-
4
682
6162781
Back To Top
More Information
Panther Classification:
Molecular Function -
phosphatase modulator
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0