Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPP6R2 SIRNA Assays ›
          Gene Symbol:
          PPP6R2
          Gene Name:
          protein phosphatase 6 regulatory subunit 2
          Gene Aliases:
          KIAA0685, PP6R2, SAP190, SAPS2
          Chromosome Location:
          Chr.22: 50343317 - 50445090 on Build GRCh38
          UniGene ID:
          Hs.449098
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s18691

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PPP6R2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242898.1 NP_001229827.1 3 484
          NM_001242899.1 NP_001229828.1 3 484
          NM_001242900.1 NP_001229829.1 3 484
          NM_014678.4 NP_055493.2 3 484
          XM_006724431.2 XP_006724494.1 4 595
          XM_006724434.1 XP_006724497.1 5 744
          XM_006724435.2 XP_006724498.1 3 469
          XM_011530720.2 XP_011529022.1 2 169
          XM_011530721.2 XP_011529023.1 2 169
          XM_011530722.2 XP_011529024.1 2 169
          XM_011530723.2 XP_011529025.1 2 169
          XM_011530724.2 XP_011529026.1 2 169
          XM_011530726.2 XP_011529028.1 2 169
          XM_011530727.2 XP_011529029.1 2 169
          XM_011530728.2 XP_011529030.1 2 169
          XM_011530729.2 XP_011529031.1 2 169
          XM_011530730.2 XP_011529032.1 2 169
          XM_011530731.1 XP_011529033.1 2 169
          XM_011530732.2 XP_011529034.1 5 645
          XM_011530734.2 XP_011529036.1 4 519
          XM_011530735.2 XP_011529037.1 3 469
          XM_011530736.2 XP_011529038.1 4 595
          XM_011530737.2 XP_011529039.1 5 744
          XM_011530738.2 XP_011529040.1 4 547
          XM_011530739.2 XP_011529041.1 2 209
          XM_011530740.2 XP_011529042.1 4 595
          XM_017029116.1 XP_016884605.1 2 169
          XM_017029117.1 XP_016884606.1 6 794
          XM_017029118.1 XP_016884607.1 6 746
          XM_017029119.1 XP_016884608.1 5 668
          XM_017029120.1 XP_016884609.1 5 645
          XM_017029121.1 XP_016884610.1 3 469
          XM_017029122.1 XP_016884611.1 4 547
          XM_017029123.1 XP_016884612.1 3 469
          XM_017029124.1 XP_016884613.1 4 595
          XM_017029125.1 XP_016884614.1 3 469
          XM_017029126.1 XP_016884615.1 3 469
          XM_017029127.1 XP_016884616.1 4 547
          XM_017029128.1 XP_016884617.1 4 595
          XM_017029129.1 XP_016884618.1 3 469
          XM_017029130.1 XP_016884619.1 3 469
          XM_017029131.1 XP_016884620.1 4 595
          XM_017029132.1 XP_016884621.1 4 547
          XM_017029133.1 XP_016884622.1 3 469
          XM_017029134.1 XP_016884623.1 3 469
          XM_017029135.1 XP_016884624.1 4 519
          XM_017029136.1 XP_016884625.1 3 469
          XM_017029137.1 XP_016884626.1 3 469
          XM_017029138.1 XP_016884627.1 3 469
          GenBank mRNA AB014585.1 - 3 456
          AK302472.1 - 3 395
          BC000976.4 - 4 473 2900437
          BC006568.2 - 3 470 3532211
          BC032664.1 - 3 481 5553142
          BC041698.1 - 3 451 5732101
          BC052995.2 - 3 471 6162781

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_027805, NM_026813, BC024879, AK018403, AK084684, and others.

          Panther Classification:

          Molecular Function -

          phosphatase modulator

          Gene Ontology Categories:

          Process(es)

          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0