Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HDAC9 SIRNA Assays ›
          Gene Symbol:
          HDAC9
          Gene Name:
          histone deacetylase 9
          Gene Aliases:
          HD7, HD7b, HD9, HDAC, HDAC7, HDAC7B, HDAC9B, HDAC9FL, HDRP, MITR
          Chromosome Location:
          Chr.7: 18086942 - 19003745 on Build GRCh38
          UniGene ID:
          Hs.196054
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s18773

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HDAC9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001204148.2 NP_001191077.1 6 849
          NM_001321869.1 NP_001308798.1 8 1131
          NM_001321870.1 NP_001308799.1 8 1122
          NM_001321874.1 NP_001308803.1 7 1052
          NM_001321875.1 NP_001308804.1 7 1058
          NM_001321876.1 NP_001308805.1 7 1046
          NM_001321878.1 NP_001308807.1 7 1043
          NM_001321884.1 NP_001308813.1 7 1046
          NM_001321886.1 NP_001308815.1 8 1102
          NM_001321887.1 NP_001308816.1 8 936
          NM_001321890.1 NP_001308819.1 7 1037
          NM_001321896.1 NP_001308825.1 8 1221
          NM_001321899.1 NP_001308828.1 8 949
          NM_001321900.1 NP_001308829.1 7 1043
          NM_001321901.1 NP_001308830.1 7 877
          NM_001321902.1 NP_001308831.1 7 883
          NM_014707.3 NP_055522.1 7 874
          NM_058176.2 NP_478056.1 6 729
          NM_178423.2 NP_848510.1 7 874
          NM_178425.3 NP_848512.1 6 738
          XM_011515629.2 XP_011513931.1 7 832
          XM_011515631.2 XP_011513933.1 8 1146
          XM_011515635.2 XP_011513937.1 8 1698
          XM_011515636.2 XP_011513938.1 7 1419
          XM_011515637.2 XP_011513939.1 7 843
          XM_011515638.2 XP_011513940.1 7 1413
          XM_011515639.2 XP_011513941.1 7 1610
          XM_011515641.2 XP_011513943.1 8 1695
          XM_011515642.2 XP_011513944.1 6 2424
          XM_011515644.2 XP_011513946.1 8 1670
          XM_011515645.2 XP_011513947.1 6 2409
          XM_011515646.2 XP_011513948.1 6 2400
          XM_011515648.2 XP_011513950.1 7 1428
          XM_011515649.2 XP_011513951.1 7 1632
          XM_017012826.1 XP_016868315.1 7 1404
          XM_017012827.1 XP_016868316.1 8 1487
          XM_017012829.1 XP_016868318.1 7 807
          XM_017012830.1 XP_016868319.1 7 798
          XM_017012832.1 XP_016868321.1 7 807
          GenBank mRNA AB018287.1 - 7 838
          AF124924.1 - 4 396
          AJ459808.1 - 7 838
          AK304343.1 - 6 769
          AY032737.1 - 6 688
          AY032738.1 - 6 688
          AY197371.1 - 6 697
          BC150328.1 - 7 849 8860495
          BC152405.1 - 7 838 8862508
          CR749550.1 - 6 805

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001200045, XM_008764616, XM_008764610, XM_008764611, AB558551, and others.
          This siRNA also targets the following transcripts in Mouse: NM_024124, NM_001271386, AF235053, AK133334, BC098187, and others.

          Panther Classification:

          Molecular Function -

          histone modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          response to amphetamine
          transcription, DNA-templated
          inflammatory response
          heart development
          chromatin modification
          histone deacetylation
          neuron differentiation
          B cell differentiation
          cellular response to insulin stimulus
          peptidyl-lysine deacetylation
          B cell activation
          negative regulation of transcription, DNA-templated
          regulation of skeletal muscle fiber development
          regulation of striated muscle cell differentiation
          histone H3 deacetylation
          histone H4 deacetylation
          positive regulation of cell migration involved in sprouting angiogenesis
          transcription corepressor activity
          histone deacetylase activity
          protein kinase C binding
          protein binding
          transcription factor binding
          NAD-dependent histone deacetylase activity (H3-K14 specific)
          protein deacetylase activity
          histone deacetylase binding
          metal ion binding
          repressing transcription factor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0