Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HDAC9 SIRNA Assays ›
          Gene Symbol:
          HDAC9
          Gene Name:
          histone deacetylase 9
          Gene Aliases:
          HD7, HD7b, HD9, HDAC, HDAC7, HDAC7B, HDAC9B, HDAC9FL, HDRP, MITR
          Chromosome Location:
          Chr.7: 18086942 - 19003745 on Build GRCh38
          UniGene ID:
          Hs.196054
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s18775

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HDAC9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001204144.2 NP_001191073.1 12 1810
          NM_001204145.2 NP_001191074.1 10 1502
          NM_001204146.2 NP_001191075.1 10 1680
          NM_001204147.2 NP_001191076.1 9 1462
          NM_001204148.2 NP_001191077.1 10 1609
          NM_001321868.1 NP_001308797.1 11 1750
          NM_001321869.1 NP_001308798.1 12 1891
          NM_001321870.1 NP_001308799.1 12 1882
          NM_001321871.1 NP_001308800.1 11 1765
          NM_001321872.1 NP_001308801.1 11 1759
          NM_001321873.1 NP_001308802.1 11 1753
          NM_001321874.1 NP_001308803.1 11 1812
          NM_001321875.1 NP_001308804.1 11 1818
          NM_001321876.1 NP_001308805.1 11 1806
          NM_001321877.1 NP_001308806.1 10 1680
          NM_001321878.1 NP_001308807.1 11 1803
          NM_001321879.1 NP_001308808.1 10 1674
          NM_001321884.1 NP_001308813.1 11 1806
          NM_001321885.1 NP_001308814.1 10 1695
          NM_001321886.1 NP_001308815.1 12 1862
          NM_001321887.1 NP_001308816.1 12 1696
          NM_001321888.1 NP_001308817.1 10 1496
          NM_001321889.1 NP_001308818.1 11 1733
          NM_001321890.1 NP_001308819.1 11 1797
          NM_001321891.1 NP_001308820.1 10 1671
          NM_001321893.1 NP_001308822.1 11 1568
          NM_001321894.1 NP_001308823.1 11 1577
          NM_001321895.1 NP_001308824.1 10 1511
          NM_001321896.1 NP_001308825.1 12 1981
          NM_001321897.1 NP_001308826.1 11 1577
          NM_001321898.1 NP_001308827.1 10 1526
          NM_001321899.1 NP_001308828.1 12 1709
          NM_001321900.1 NP_001308829.1 11 1803
          NM_001321901.1 NP_001308830.1 11 1637
          NM_001321902.1 NP_001308831.1 11 1643
          NM_014707.3 NP_055522.1 11 1634
          NM_058176.2 NP_478056.1 10 1489
          NM_178423.2 NP_848510.1 11 1634
          NM_178425.3 NP_848512.1 10 1498
          XM_011515629.2 XP_011513931.1 11 1592
          XM_011515631.2 XP_011513933.1 12 1906
          XM_011515634.2 XP_011513936.1 10 1460
          XM_011515635.2 XP_011513937.1 12 2458
          XM_011515636.2 XP_011513938.1 11 2179
          XM_011515637.2 XP_011513939.1 11 1603
          XM_011515638.2 XP_011513940.1 11 2173
          XM_011515639.2 XP_011513941.1 11 2370
          XM_011515641.2 XP_011513943.1 12 2455
          XM_011515642.2 XP_011513944.1 10 3184
          XM_011515644.2 XP_011513946.1 12 2430
          XM_011515645.2 XP_011513947.1 10 3169
          XM_011515646.2 XP_011513948.1 10 3160
          XM_011515648.2 XP_011513950.1 11 2188
          XM_011515649.2 XP_011513951.1 11 2392
          XM_011515651.1 XP_011513953.1 10 1426
          XM_017012826.1 XP_016868315.1 11 2164
          XM_017012827.1 XP_016868316.1 12 2247
          XM_017012828.1 XP_016868317.1 10 2240
          XM_017012829.1 XP_016868318.1 11 1567
          XM_017012830.1 XP_016868319.1 11 1558
          XM_017012831.1 XP_016868320.1 10 1435
          XM_017012832.1 XP_016868321.1 11 1567
          GenBank mRNA AB018287.1 - 11 1598
          AF124924.1 - 8 1156
          AJ459808.1 - 11 1598
          AK097178.1 - 10 1460
          AK296214.1 - 10 1412
          AK297404.1 - 10 1456
          AK304298.1 - 12 1803
          AK304343.1 - 10 1529
          AK304410.1 - 9 1425
          AK316109.1 - 10 1631
          AY032737.1 - 10 1448
          AY032738.1 - 10 1448
          AY197371.1 - 10 1457
          BC111735.1 - 9 1324
          BC150328.1 - 11 1609 8860495
          BC152405.1 - 11 1598 8862508
          CR749550.1 - 10 1565

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001200045, XM_008764615, XM_008764610, XM_008764613, XM_008764616, and others.
          This siRNA also targets the following transcripts in Mouse: NM_024124, NM_001271386, AK144242, AF235053, AK133334, and others.

          Panther Classification:

          Molecular Function -

          histone modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          response to amphetamine
          transcription, DNA-templated
          inflammatory response
          heart development
          chromatin modification
          histone deacetylation
          neuron differentiation
          B cell differentiation
          cellular response to insulin stimulus
          peptidyl-lysine deacetylation
          B cell activation
          negative regulation of transcription, DNA-templated
          regulation of skeletal muscle fiber development
          regulation of striated muscle cell differentiation
          histone H3 deacetylation
          histone H4 deacetylation
          positive regulation of cell migration involved in sprouting angiogenesis
          transcription corepressor activity
          histone deacetylase activity
          protein kinase C binding
          protein binding
          transcription factor binding
          NAD-dependent histone deacetylase activity (H3-K14 specific)
          protein deacetylase activity
          histone deacetylase binding
          metal ion binding
          repressing transcription factor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0