Search
Search

Pramel1| Gene Symbol: Pramel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109617.1 | NP_001103087.1 | 1261 | |||
| XM_006239342.2 | XP_006239404.1 | 4 | 1404 | |||
| XM_006239343.2 | XP_006239405.1 | 5 | 1571 | |||
| XM_008764290.1 | XP_008762512.1 | 1413 | ||||