Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LOC100361558 SIRNA Assays ›
          Gene Symbol:
          LOC100361558 H3f3b H3f3c LOC100359421
          Gene Name:
          histone H3.3B-like
          H3 histone, family 3B
          H3 histone, family 3C
          histone H3.3B-like
          Gene Aliases:
          -
          H3.3B
          -
          -
          Chromosome Location:
          Chr.13: 99091246 - 99102828 on Build Rnor_6.0
          Chromosome Location:
          Chr.10: 104573666 - 104575890 on Build Rnor_6.0
          Chromosome Location:
          Chr.7: 94777648 - 94778697 on Build Rnor_6.0
          Chromosome Location:
          Chr.6: 60988487 - 60989918 on Build Rnor_6.0
          UniGene ID:
          Rn.106155
          Rn.124815
          -
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          siRNA ID s189582

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LOC100361558
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_002728043.3 XP_002728089.1 4 916
          GenBank mRNA FQ209679.1 - 1 405
          FQ212662.1 - 1 404
          FQ212848.1 - 1 405
          FQ213184.1 - 1 404
          FQ213303.1 - 1 404
          FQ213733.1 - 1 404
          FQ213904.1 - 1 404
          FQ214463.1 - 1 404
          FQ215011.1 - 1 404
          FQ215494.1 - 1 404
          FQ216000.1 - 1 405
          FQ216532.1 - 1 404
          FQ216541.1 - 1 405
          FQ217113.1 - 1 404
          FQ218544.1 - 1 404
          FQ218621.1 - 1 404
          FQ218899.1 - 1 404
          FQ219801.1 - 1 404
          FQ220103.1 - 1 405
          FQ220271.1 - 1 403
          FQ220394.1 - 1 404
          FQ220658.1 - 1 404
          FQ220729.1 - 1 404
          FQ224168.1 - 1 404
          FQ224592.1 - 1 403
          FQ224944.1 - 1 404
          FQ226485.1 - 1 404
          FQ227494.1 - 1 404
          FQ227698.1 - 1 404
          FQ228409.1 - 1 404
          FQ228471.1 - 1 404
          FQ229363.1 - 1 404
          FQ229473.1 - 1 404
          FQ229495.1 - 1 404
          FQ229510.1 - 1 404
          FQ229613.1 - 1 404
          FQ229729.1 - 1 404
          FQ229838.1 - 1 404
          FQ229878.1 - 1 404
          FQ229962.1 - 1 404
          FQ229981.1 - 1 404
          FQ230073.1 - 1 405
          FQ230430.1 - 1 404
          FQ232416.1 - 1 404
          FQ232524.1 - 1 404
          FQ233003.1 - 1 404
          FQ234024.1 - 1 404
          FQ235248.1 - 1 404
          Gene Symbol: H3f3b
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC086580.1 - 878 7307209
          BC087725.1 - 848 7320825
          BC159439.1 - 880 9024967
          Gene Symbol: H3f3c
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_003750348.3 XP_003750396.1 1 902
          Gene Symbol: LOC100359421
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_002729644.4 XP_002729690.2 2 838

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          nucleosome assembly
          DNA replication-independent nucleosome assembly
          biological_process
          positive regulation of cell growth
          muscle cell differentiation
          negative regulation of chromosome condensation
          RNA polymerase II core promoter sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          molecular_function
          nucleosomal DNA binding
          DNA binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0