Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SMG7 SIRNA Assays ›
Gene Symbol:
SMG7
Gene Name:
SMG7, nonsense mediated mRNA decay factor
Gene Aliases:
C1orf16, EST1C, SGA56M
Chromosome Location:
Chr.1: 183471962 - 183554193 on Build GRCh38
UniGene ID:
Hs.591463
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s19152
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SMG7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001174061.1
NP_001167532.1
16
2857
NM_173156.2
NP_775179.1
17
2889
NM_201568.2
NP_963862.1
17
2751
NM_201569.2
NP_963863.2
17
2751
XM_005245648.3
XP_005245705.1
19
3574
XM_005245649.3
XP_005245706.1
18
3437
XM_005245652.3
XP_005245709.1
17
2758
XM_005245653.4
XP_005245710.1
18
2860
XM_006711676.3
XP_006711739.1
18
3299
XM_006711678.3
XP_006711741.1
17
2620
XM_011510205.2
XP_011508507.1
20
3752
XM_011510206.2
XP_011508508.1
20
3614
XM_011510207.2
XP_011508509.1
18
3476
XM_011510208.2
XP_011508510.1
20
3752
XM_011510209.2
XP_011508511.1
20
3614
XM_011510210.2
XP_011508512.1
16
2590
XM_017002966.1
XP_016858455.1
19
2891
XM_017002967.1
XP_016858456.1
19
2899
XM_017002968.1
XP_016858457.1
16
2594
XM_017002969.1
XP_016858458.1
17
2951
XM_017002970.1
XP_016858459.1
17
2875
XM_017002971.1
XP_016858460.1
17
2813
GenBank mRNA
AB085674.1
-
17
2702
AK299178.1
-
16
2728
AL833675.1
-
5
1134
AY386363.1
-
10
1858
BC036381.1
-
17
2619
4829267
BC052565.1
-
10
1746
D87437.2
-
17
2595
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
mRNA export from nucleus
regulation of dephosphorylation
protein binding
telomeric DNA binding
protein phosphatase 2A binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0