Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SNAP91 SIRNA Assays ›
Gene Symbol:
SNAP91
Gene Name:
synaptosome associated protein 91
Gene Aliases:
AP180, CALM
Chromosome Location:
Chr.6: 83552885 - 83709408 on Build GRCh38
UniGene ID:
Hs.368046
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s19161
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SNAP91
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242792.1
NP_001229721.1
25
2627
NM_001242793.1
NP_001229722.1
23
2336
NM_001242794.1
NP_001229723.1
15
1505
NM_001256717.1
NP_001243646.1
25
2694
NM_001256718.1
NP_001243647.1
24
2322
NM_014841.2
NP_055656.1
25
2630
XM_005248770.4
XP_005248827.1
25
2469
XM_006715615.1
XP_006715678.1
25
2421
XM_011536265.1
XP_011534567.1
25
2428
XM_011536266.1
XP_011534568.1
25
2540
XM_011536269.1
XP_011534571.1
25
2406
XM_011536271.1
XP_011534573.1
24
2379
XM_011536273.2
XP_011534575.1
24
2343
XM_011536275.2
XP_011534577.1
24
2337
XM_011536276.2
XP_011534578.1
24
2313
XM_011536277.1
XP_011534579.1
27
2709
XM_017011554.1
XP_016867043.1
24
2579
XM_017011555.1
XP_016867044.1
24
2463
XM_017011556.1
XP_016867045.1
24
2395
XM_017011557.1
XP_016867046.1
25
2583
XM_017011558.1
XP_016867047.1
25
2413
XM_017011559.1
XP_016867048.1
25
2512
XM_017011560.1
XP_016867049.1
25
2454
XM_017011561.1
XP_016867050.1
25
2525
XM_017011562.1
XP_016867051.1
24
2583
XM_017011563.1
XP_016867052.1
23
2577
XM_017011564.1
XP_016867053.1
23
2448
XM_017011565.1
XP_016867054.1
23
2506
XM_017011566.1
XP_016867055.1
24
2556
XM_017011567.1
XP_016867056.1
24
2359
XM_017011568.1
XP_016867057.1
23
2550
XM_017011569.1
XP_016867058.1
24
2541
XM_017011570.1
XP_016867059.1
24
2520
XM_017011571.1
XP_016867060.1
24
2391
XM_017011572.1
XP_016867061.1
24
2350
XM_017011573.1
XP_016867062.1
24
2449
XM_017011574.1
XP_016867063.1
25
2598
XM_017011575.1
XP_016867064.1
24
2514
XM_017011576.1
XP_016867065.1
24
2317
XM_017011577.1
XP_016867066.1
24
2443
XM_017011578.1
XP_016867067.1
24
2456
XM_017011579.1
XP_016867068.1
24
2499
XM_017011580.1
XP_016867069.1
24
2490
XM_017011581.1
XP_016867070.1
24
2320
XM_017011582.1
XP_016867071.1
24
2349
XM_017011583.1
XP_016867072.1
24
2462
XM_017011584.1
XP_016867073.1
24
2305
XM_017011585.1
XP_016867074.1
24
2346
XM_017011586.1
XP_016867075.1
24
2298
XM_017011587.1
XP_016867076.1
24
2404
XM_017011588.1
XP_016867077.1
24
2417
XM_017011589.1
XP_016867078.1
23
2448
XM_017011590.1
XP_016867079.1
23
2277
GenBank mRNA
AB014556.1
-
25
2556
AK289582.1
-
25
2598
AK294556.1
-
18
1588
AK294898.1
-
24
2306
BC060818.1
-
15
1489
4827211
CR749348.1
-
25
2661
Back To Top
More Information
Panther Classification:
Molecular Function -
vesicle coat protein
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
protein transport
clathrin coat assembly
regulation of clathrin-mediated endocytosis
protein binding
1-phosphatidylinositol binding
protein kinase binding
clathrin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0