Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ABI1 SIRNA Assays ›
          Gene Symbol:
          ABI1
          Gene Name:
          abl interactor 1
          Gene Aliases:
          ABI-1, ABLBP4, E3B1, NAP1BP, SSH3BP, SSH3BP1
          Chromosome Location:
          Chr.10: 26746596 - 26861087 on Build GRCh38
          UniGene ID:
          Hs.508148
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s19444

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ABI1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001012750.2 NP_001012768.1 2 402
          NM_001012751.2 NP_001012769.1 2 402
          NM_001012752.2 NP_001012770.1 2 402
          NM_001178116.1 NP_001171587.1 3 453
          NM_001178119.1 NP_001171590.1 2 402
          NM_001178120.1 NP_001171591.1 2 402
          NM_001178121.1 NP_001171592.1 2 402
          NM_001178122.1 NP_001171593.1 2 402
          NM_001178123.1 NP_001171594.1 2 402
          NM_001178124.1 NP_001171595.1 2 402
          NM_001178125.1 NP_001171596.1 2 402
          NM_005470.3 NP_005461.2 2 402
          XM_005252332.3 XP_005252389.1 2 217
          XM_005252333.3 XP_005252390.1 2 217
          XM_006717359.2 XP_006717422.1 2 301
          XM_006717360.2 XP_006717423.1 2 301
          XM_006717361.2 XP_006717424.1 2 217
          XM_011519289.2 XP_011517591.1 2 301
          XM_017015458.1 XP_016870947.1 2 301
          XM_017015459.1 XP_016870948.1 2 301
          GenBank mRNA AB040151.1 - 2 345
          AB209268.1 - 2 253
          AF001628.1 - 2 226
          AF006516.1 - 2 244
          AF260262.1 - 2 301
          AF540955.1 - 2 190
          AK126803.1 - 3 352
          AK291823.1 - 2 263
          AK298646.1 - 2 299
          BC013238.1 - 2 250
          BC024254.2 - 2 253 3531592
          DB019974.1 - 2 403
          FJ380057.1 - 2 178
          FJ380058.1 - 2 178
          FJ380059.1 - 2 178
          U87166.1 - 2 259

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_024397, XM_006254356, XM_006254353, XM_006254354, XM_006254357, and others.
          This siRNA also targets the following transcripts in Mouse: NM_145994, NM_001077190, NM_001077192, NM_007380, NM_001077193, and others.

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          somitogenesis
          movement of cell or subcellular component
          transmembrane receptor protein tyrosine kinase signaling pathway
          actin polymerization or depolymerization
          negative regulation of cell proliferation
          viral process
          peptidyl-tyrosine phosphorylation
          megakaryocyte development
          Fc-gamma receptor signaling pathway involved in phagocytosis
          vascular endothelial growth factor receptor signaling pathway
          positive regulation of protein tyrosine kinase activity
          lamellipodium morphogenesis
          cell-cell adhesion
          protein binding
          cytoskeletal protein binding
          protein tyrosine kinase activator activity
          protein complex binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-d8jzh:80/100.66.76.85:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0