Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HMOX1 SIRNA Assays ›
Gene Symbol:
HMOX1
Gene Name:
heme oxygenase 1
Gene Aliases:
HMOX1D, HO-1, HSP32, bK286B10
Chromosome Location:
Chr.22: 35381067 - 35394214 on Build GRCh38
UniGene ID:
Hs.517581
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s194530
Validated
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
7,898.00
Your Price
Online offer:
7898.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HMOX1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002133.2
NP_002124.1
5
1438
GenBank mRNA
BC001491.2
-
5
1397
3504480
M23042.1
-
1
365
X06985.1
-
5
1411
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00157965_m1
Panther Classification:
Molecular Function -
oxygenase
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
endothelial cell proliferation
wound healing involved in inflammatory response
negative regulation of leukocyte migration
heme oxidation
cellular iron ion homeostasis
response to oxidative stress
small GTPase mediated signal transduction
excretion
regulation of blood pressure
cell death
intrinsic apoptotic signaling pathway in response to DNA damage
negative regulation of muscle cell apoptotic process
smooth muscle hyperplasia
cellular response to nutrient
negative regulation of mast cell cytokine production
erythrocyte homeostasis
low-density lipoprotein particle clearance
regulation of transcription from RNA polymerase II promoter in response to iron
cellular response to heat
response to nicotine
intracellular signal transduction
heme catabolic process
response to hydrogen peroxide
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of mast cell degranulation
negative regulation of DNA binding
negative regulation of sequence-specific DNA binding transcription factor activity
negative regulation of neuron apoptotic process
regulation of transcription from RNA polymerase II promoter in response to oxidative stress
response to estrogen
positive regulation of chemokine biosynthetic process
regulation of angiogenesis
positive regulation of angiogenesis
positive regulation of vasodilation
positive regulation of smooth muscle cell proliferation
negative regulation of smooth muscle cell proliferation
regulation of sequence-specific DNA binding transcription factor activity
protein homooligomerization
iron ion homeostasis
cellular response to arsenic-containing substance
cellular response to cadmium ion
cellular response to hypoxia
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
heme oxygenase (decyclizing) activity
phospholipase D activity
signal transducer activity
protein binding
enzyme binding
heme binding
protein homodimerization activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0