Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SCNN1D SIRNA Assays ›
Gene Symbol:
SCNN1D
Gene Name:
sodium channel epithelial 1 delta subunit
Gene Aliases:
ENaCd, ENaCdelta, SCNED, dNaCh
Chromosome Location:
Chr.1: 1280415 - 1292029 on Build GRCh38
UniGene ID:
Hs.512681
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s194785
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SCNN1D
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130413.3
NP_001123885.2
11
1697
NR_037668.2
10
1603
XM_011541899.2
XP_011540201.1
11
2367
XM_011541901.2
XP_011540203.1
12
1889
XM_011541902.2
XP_011540204.1
12
1865
XM_011541905.2
XP_011540207.1
11
1793
XM_011541906.2
XP_011540208.1
8
2127
XM_011541908.2
XP_011540210.1
11
2064
XM_011541920.2
XP_011540222.1
10
1887
XM_011541925.2
XP_011540227.1
9
1446
XM_011541929.2
XP_011540231.1
7
2255
XM_011541932.2
XP_011540234.1
8
1352
XM_011541933.2
XP_011540235.1
8
1318
XM_017002037.1
XP_016857526.1
10
2364
XM_017002038.1
XP_016857527.1
9
2217
XM_017002039.1
XP_016857528.1
11
1742
XM_017002040.1
XP_016857529.1
11
1604
XM_017002041.1
XP_016857530.1
12
1592
XM_017002042.1
XP_016857531.1
11
1583
XM_017002043.1
XP_016857532.1
11
1559
XM_017002044.1
XP_016857533.1
10
1508
GenBank mRNA
AK093239.1
-
8
1327
AK093372.1
-
5
452
AK127357.1
-
8
2123
AK131558.1
-
12
2023
AK315288.1
-
7
1174
BC036752.1
-
7
1146
BC040882.1
-
10
1567
BC125074.1
-
7
1096
40117641
BC125075.1
-
7
1096
40117643
DQ898175.1
-
7
1071
DQ898176.1
-
8
1177
EU489064.1
-
6
979
U38254.1
-
6
979
Back To Top
More Information
Panther Classification:
Molecular Function -
ion channel
Gene Ontology Categories:
Function(s)
Process(es)
sodium ion transport
ion transmembrane transport
sodium ion transmembrane transport
response to stimulus
sensory perception of taste
protein binding
ligand-gated sodium channel activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline