Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ECHDC2 SIRNA Assays ›
Gene Symbol:
ECHDC2
Gene Name:
enoyl-CoA hydratase domain containing 2
Gene Aliases:
-
Chromosome Location:
Chr.1: 52895910 - 52927212 on Build GRCh38
UniGene ID:
Hs.476319
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s195278
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ECHDC2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198961.1
NP_001185890.1
9
879
NM_001198962.1
NP_001185891.1
7
735
NM_001319958.1
NP_001306887.1
9
1299
NM_018281.3
NP_060751.2
8
786
XM_011541709.2
XP_011540011.1
11
1265
XM_011541710.2
XP_011540012.1
10
1090
XM_011541711.2
XP_011540013.1
10
1172
XM_011541713.2
XP_011540015.1
10
5042
XM_011541715.2
XP_011540017.1
11
1490
XM_011541719.2
XP_011540021.1
11
2487
XM_011541720.2
XP_011540022.1
10
4811
XM_011541721.2
XP_011540023.1
12
1504
XM_011541722.2
XP_011540024.1
11
1122
XM_011541723.2
XP_011540025.1
11
1222
XM_011541724.2
XP_011540026.1
9
1115
XM_011541725.2
XP_011540027.1
9
997
XM_011541726.2
XP_011540028.1
9
4949
XM_011541727.2
XP_011540029.1
8
4661
XM_017001638.1
XP_016857127.1
12
2876
XM_017001639.1
XP_016857128.1
11
1281
XM_017001640.1
XP_016857129.1
10
1444
XM_017001641.1
XP_016857130.1
12
2645
XM_017001642.1
XP_016857131.1
8
4774
GenBank mRNA
AF258590.1
-
4
1698
AK001810.1
-
8
738
AK302285.1
-
12
1458
AL832874.1
-
2
1947
BC038389.1
-
9
1217
BC044574.1
-
9
809
5314083
BX647186.1
-
1
4578
CF127686.1
-
7
679
Back To Top
More Information
Panther Classification:
Molecular Function -
hydratase
Gene Ontology Categories:
Function(s)
Process(es)
fatty acid beta-oxidation
enoyl-CoA hydratase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0