Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BDNF SIRNA Assays ›
          Gene Symbol:
          BDNF
          Gene Name:
          brain derived neurotrophic factor
          Gene Aliases:
          ANON2, BULN2
          Chromosome Location:
          Chr.11: 27654893 - 27722058 on Build GRCh38
          UniGene ID:
          Hs.502182
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s1962

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BDNF
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001143805.1 NP_001137277.1 2 182
          NM_001143806.1 NP_001137278.1 2 397
          NM_001143807.1 NP_001137279.1 2 283
          NM_001143808.1 NP_001137280.1 2 119
          NM_001143809.1 NP_001137281.1 2 129
          NM_001143810.1 NP_001137282.1 3 412
          NM_001143811.1 NP_001137283.1 4 529
          NM_001143812.1 NP_001137284.1 2 271
          NM_001143813.1 NP_001137285.1 2 353
          NM_001143814.1 NP_001137286.1 3 478
          NM_001143816.1 NP_001137288.1 2 876
          NM_001709.4 NP_001700.2 2 371
          NM_170731.4 NP_733927.1 2 693
          NM_170732.4 NP_733928.1 2 480
          NM_170733.3 NP_733929.1 2 383
          NM_170734.3 NP_733930.1 2 331
          NM_170735.5 NP_733931.1 1 1110
          XM_011520280.2 XP_011518582.1 2 1277
          GenBank mRNA AB038670.1 - 2 363
          AB038671.1 - 2 380
          AB038672.1 - 2 265
          AB038673.1 - 2 100
          AB038674.1 - 2 298
          AF400438.1 - 1 43
          AK289763.1 - 2 179
          AK289853.1 - 2 80
          AY054392.2 - 2 384
          AY054393.2 - 2 126
          AY054394.2 - 2 341
          AY054395.2 - 2 424
          AY054396.2 - 2 182
          AY054397.2 - 2 265
          AY054398.2 - 2 312
          AY054399.2 - 3 197
          AY054400.2 - 2 298
          AY054406.3 - 2 166
          AY442188.1 - 2 194
          AY656701.1 - 1 25
          BC029795.1 - 2 356 5193877
          CX751420.1 - 2 250
          DQ250642.1 - 2 112
          EF674517.1 - 2 182
          EF674518.1 - 2 397
          EF674519.1 - 2 480
          EF674520.1 - 2 283
          EF674521.1 - 2 383
          EF689009.1 - 1 1124
          EF689010.1 - 2 890
          EF689011.1 - 2 119
          EF689012.1 - 2 129
          EF689013.1 - 2 271
          EF689014.1 - 2 353
          EF689015.1 - 2 371
          EF689016.1 - 3 478
          EF689017.1 - 3 230
          EF689018.1 - 2 331
          EF689019.1 - 3 412
          EF689020.1 - 4 529
          EF689021.1 - 2 693
          KP229412.1 - 1 25
          KU177943.1 - 1 25
          KU177944.1 - 1 25
          KU177945.1 - 1 25
          M61176.1 - 1 587
          X60201.1 - 2 321
          X91251.1 - 2 309

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001270633, NM_012513, NM_001270630, NM_001270631, NM_001270638, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001285420, NM_001285419, NM_001285417, NM_001285421, NM_001048139, and others.

          Panther Classification:

          Molecular Function -

          neurotrophic factor

          Gene Ontology Categories:

          Function(s) Process(es)

          cell-cell signaling
          nervous system development
          axon guidance
          synapse assembly
          brain-derived neurotrophic factor receptor signaling pathway
          positive regulation of brain-derived neurotrophic factor receptor signaling pathway
          negative regulation of neuron apoptotic process
          collateral sprouting
          positive regulation of collateral sprouting
          positive regulation of synapse assembly
          neurotrophin TRKB receptor binding
          growth factor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0