Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BDNF SIRNA Assays ›
Gene Symbol:
BDNF
Gene Name:
brain derived neurotrophic factor
Gene Aliases:
ANON2, BULN2
Chromosome Location:
Chr.11: 27654893 - 27722058 on Build GRCh38
UniGene ID:
Hs.502182
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s1963
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BDNF
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001143805.1
NP_001137277.1
2
443
NM_001143806.1
NP_001137278.1
2
658
NM_001143807.1
NP_001137279.1
2
544
NM_001143808.1
NP_001137280.1
2
380
NM_001143809.1
NP_001137281.1
2
390
NM_001143810.1
NP_001137282.1
3
673
NM_001143811.1
NP_001137283.1
4
790
NM_001143812.1
NP_001137284.1
2
532
NM_001143813.1
NP_001137285.1
2
614
NM_001143814.1
NP_001137286.1
3
739
NM_001143816.1
NP_001137288.1
2
1137
NM_001709.4
NP_001700.2
2
632
NM_170731.4
NP_733927.1
2
954
NM_170732.4
NP_733928.1
2
741
NM_170733.3
NP_733929.1
2
644
NM_170734.3
NP_733930.1
2
592
NM_170735.5
NP_733931.1
1
1371
XM_011520280.2
XP_011518582.1
2
1538
GenBank mRNA
AF400438.1
-
1
304
AK289763.1
-
2
440
AK289853.1
-
2
341
AY054392.2
-
2
645
AY054393.2
-
2
387
AY054394.2
-
2
602
AY054395.2
-
2
685
AY054396.2
-
2
443
AY054397.2
-
2
526
AY054398.2
-
2
573
AY054399.2
-
3
458
AY054400.2
-
2
559
AY054406.3
-
2
427
AY656701.1
-
1
286
BC029795.1
-
2
617
5193877
CX751420.1
-
2
511
EF674517.1
-
2
443
EF674518.1
-
2
658
EF674519.1
-
2
741
EF674520.1
-
2
544
EF674521.1
-
2
644
EF689009.1
-
1
1385
EF689010.1
-
2
1151
EF689011.1
-
2
380
EF689012.1
-
2
390
EF689013.1
-
2
532
EF689014.1
-
2
614
EF689015.1
-
2
632
EF689016.1
-
3
739
EF689017.1
-
3
491
EF689018.1
-
2
592
EF689019.1
-
3
673
EF689020.1
-
4
790
EF689021.1
-
2
954
KP229412.1
-
1
286
KU177943.1
-
1
286
KU177944.1
-
1
286
KU177945.1
-
1
286
M61176.1
-
1
848
X60201.1
-
2
582
X91251.1
-
2
570
Back To Top
More Information
Panther Classification:
Molecular Function -
neurotrophic factor
Gene Ontology Categories:
Function(s)
Process(es)
cell-cell signaling
nervous system development
axon guidance
synapse assembly
brain-derived neurotrophic factor receptor signaling pathway
positive regulation of brain-derived neurotrophic factor receptor signaling pathway
negative regulation of neuron apoptotic process
collateral sprouting
positive regulation of collateral sprouting
positive regulation of synapse assembly
neurotrophin TRKB receptor binding
growth factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0