Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ABI2 SIRNA Assays ›
          Gene Symbol:
          ABI2
          Gene Name:
          abl-interactor 2
          Gene Aliases:
          ABI-2, ABI2B, AIP-1, AblBP3, SSH3BP2, argBP1, argBPIA, argBPIB
          Chromosome Location:
          Chr.2: 203327874 - 203432173 on Build GRCh38
          UniGene ID:
          Hs.471156
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s19770

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ABI2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282925.1 NP_001269854.1 2, 3 571
          NM_001282926.1 NP_001269855.1 2, 3 571
          NM_005759.5 NP_005750.4 2, 3 571
          XM_005246217.2 XP_005246274.1 2, 3 936
          XM_005246218.2 XP_005246275.1 2, 3 936
          XM_005246221.2 XP_005246278.1 2, 3 936
          XM_005246225.2 XP_005246282.1 2, 3 936
          XM_005246226.2 XP_005246283.1 2, 3 936
          XM_006712174.2 XP_006712237.1 2, 3 936
          XM_006712179.2 XP_006712242.1 2, 3 936
          XM_006712180.2 XP_006712243.1 2, 3 936
          XM_006712181.2 XP_006712244.1 2, 3 936
          XM_006712182.2 XP_006712245.1 2, 3 936
          XM_006712186.2 XP_006712249.1 2, 3 936
          XM_011510472.2 XP_011508774.1 2, 3 936
          XM_011510473.2 XP_011508775.1 1, 2 178
          XM_017003118.1 XP_016858607.1 2, 3 936
          XM_017003119.1 XP_016858608.1 2, 3 936
          XM_017003120.1 XP_016858609.1 2, 3 936
          XM_017003121.1 XP_016858610.1 2, 3 936
          XM_017003122.1 XP_016858611.1 2, 3 936
          XM_017003123.1 XP_016858612.1 2, 3 936
          XM_017003124.1 XP_016858613.1 2, 3 936
          XM_017003125.1 XP_016858614.1 2, 3 936
          XM_017003126.1 XP_016858615.1 2, 3 936
          XM_017003127.1 XP_016858616.1 2, 3 936
          XM_017003129.1 XP_016858618.1 2, 3 936
          XM_017003130.1 XP_016858619.1 2, 3 936
          XM_017003131.1 XP_016858620.1 2, 3 936
          XM_017003132.1 XP_016858621.1 2, 3 936
          XM_017003134.1 XP_016858623.1 2, 3 936
          XM_017003135.1 XP_016858624.1 2, 3 936
          XM_017003136.1 XP_016858625.1 2, 3 936
          XM_017003138.1 XP_016858627.1 2, 3 936
          XM_017003141.1 XP_016858630.1 2, 3 936
          XM_017003142.1 XP_016858631.1 2, 3 936
          XM_017003144.1 XP_016858633.1 2, 3 936
          XM_017003145.1 XP_016858634.1 2, 3 936
          XM_017003146.1 XP_016858635.1 2, 3 936
          GenBank mRNA AF260261.1 - 2, 3 295
          AK299824.1 - 2, 3 415
          BC001439.2 - 2, 3 320 3138341
          BT009920.1 - 2, 3 295
          DC376086.1 - 2, 3 420
          U31089.1 - 1, 2 40
          X95632.1 - 2, 3 395

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_173143, BC097273, FQ222350, U94904.
          This siRNA also targets the following transcripts in Mouse: NM_001198571, NM_001198570, NM_198127, AK081803, BC079545, and others.

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          movement of cell or subcellular component
          cytoskeleton organization
          actin polymerization or depolymerization
          viral process
          cell migration
          Rac protein signal transduction
          peptidyl-tyrosine phosphorylation
          positive regulation of Arp2/3 complex-mediated actin nucleation
          DNA binding
          protein binding
          cytoskeletal adaptor activity
          SH3 domain binding
          kinase binding
          ubiquitin protein ligase binding
          protein complex binding
          Rac GTPase binding
          proline-rich region binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0