Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other D2HGDH SIRNA Assays ›
Gene Symbol:
D2HGDH
Gene Name:
D-2-hydroxyglutarate dehydrogenase
Gene Aliases:
D2HGD
Chromosome Location:
Chr.2: 241734579 - 241768816 on Build GRCh38
UniGene ID:
Hs.516813
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s198464
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: D2HGDH
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001287249.1
NP_001274178.1
3
195
NM_152783.4
NP_689996.4
4
579
NR_109778.1
4
579
XM_011511734.1
XP_011510036.1
4
530
XM_011511735.1
XP_011510037.1
4
530
XM_011511736.1
XP_011510038.1
4
530
XM_011511737.2
XP_011510039.1
4
530
XM_011511738.2
XP_011510040.1
4
530
XM_011511739.1
XP_011510041.1
4
530
XM_011511740.2
XP_011510042.1
4
530
XM_011511741.2
XP_011510043.1
4
530
XM_011511743.1
XP_011510045.1
4
530
XM_011511744.1
XP_011510046.1
4
530
XM_011511745.2
XP_011510047.1
4
530
XM_011511746.2
XP_011510048.1
4
530
XM_011511747.2
XP_011510049.1
4
530
XM_011511749.2
XP_011510051.1
4
530
XM_011511750.2
XP_011510052.1
4
530
XM_011511751.1
XP_011510053.1
4
530
XM_011511752.1
XP_011510054.1
4
530
XM_011511753.2
XP_011510055.1
4
530
XM_011511756.1
XP_011510058.1
4
530
XM_011511757.2
XP_011510059.1
4
530
XM_011511758.2
XP_011510060.1
4
530
XM_011511759.2
XP_011510061.1
4
530
XM_011511760.2
XP_011510062.1
4
530
XM_017004827.1
XP_016860316.1
4
530
XM_017004828.1
XP_016860317.1
4
530
XM_017004829.1
XP_016860318.1
4
530
XM_017004830.1
XP_016860319.1
4
530
XM_017004831.1
XP_016860320.1
3
418
XM_017004832.1
XP_016860321.1
4
530
GenBank mRNA
AK091725.1
-
4
528
AK094149.1
-
4
172
AK304764.1
-
4
579
AK304773.1
-
4
528
BC036604.2
-
4
518
5274813
CB997999.1
-
4
569
Back To Top
More Information
Panther Classification:
Molecular Function -
dehydrogenase
oxidoreductase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
2-oxoglutarate metabolic process
response to manganese ion
response to zinc ion
response to cobalt ion
response to magnesium ion
cellular protein metabolic process
response to calcium ion
oxidation-reduction process
flavin adenine dinucleotide binding
(R)-2-hydroxyglutarate dehydrogenase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0