Search
Search

Prr5| Gene Symbol: Prr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146061.4 | NP_666173.4 | 7 | 810 | ||
| GenBank mRNA | AF482997.1 | - | 7 | 805 | ||
| AK046057.1 | - | 8 | 627 | |||
| AK081231.1 | - | 7 | 806 | |||
| BC024991.1 | - | 8 | 694 | 5065129 | ||