Search
Search

Ctgf| Gene Symbol: Ctgf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010217.2 | NP_034347.2 | 4 | 958 | ||
| GenBank mRNA | BC006783.1 | - | 4 | 879 | 3589136 | |
| BQ892143.1 | - | 1 | 95 | |||
| CJ055589.1 | - | 2 | 337 | |||
| M70642.1 | - | 4 | 811 | |||
| M80263.1 | - | 4 | 877 | |||