Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLEC SIRNA Assays ›
Gene Symbol:
Plec
Gene Name:
plectin
Gene Aliases:
AA591047, AU042537, EBS1, PCN, PLTN, Plec1
Chromosome Location:
Chr.15: 76170974 - 76231378 on Build GRCm38
UniGene ID:
Mm.234912
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s201801
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Plec
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001163540.1
NP_001157012.1
17
2557
NM_001163542.1
NP_001157014.1
18
2074
NM_001163549.1
NP_001157021.1
17
2120
NM_001164203.1
NP_001157675.1
18
2061
NM_011117.2
NP_035247.2
16
2023
NM_201385.2
NP_958787.2
16
1958
NM_201386.2
NP_958788.2
16
2019
NM_201387.3
NP_958789.3
16
2048
NM_201388.2
NP_958790.2
16
2010
NM_201389.2
NP_958791.2
16
2542
NM_201390.2
NP_958792.2
16
1957
NM_201391.2
NP_958793.2
16
2105
NM_201392.2
NP_958794.2
16
1931
NM_201393.2
NP_958795.2
16
2048
NM_201394.2
NP_958796.2
16
2105
GenBank mRNA
AF188008.1
-
17
2557
AF188011.1
-
17
2120
AF188012.1
-
16
2105
AF188013.1
-
18
2074
AF188014.1
-
16
2023
AF188021.1
-
16
1931
AF188023.1
-
18
2061
AK017743.1
-
16
1938
AY480033.1
-
16
2023
AY480034.1
-
16
1958
AY480035.1
-
16
2019
AY480036.1
-
16
2048
AY480037.1
-
16
2010
AY480038.1
-
16
2542
AY480039.1
-
16
1957
AY480040.1
-
16
2105
AY480041.1
-
16
1931
AY480042.1
-
16
2048
AY480043.1
-
16
2105
BC021810.1
-
16
2092
BC099929.1
-
16
2543
Back To Top
More Information
Panther Classification:
Molecular Function -
intermediate filament binding protein
Gene Ontology Categories:
Function(s)
Process(es)
response to nutrient
hemidesmosome assembly
actin binding
structural constituent of cytoskeleton
protein binding
cytoskeletal protein binding
structural constituent of muscle
ankyrin binding
poly(A) RNA binding
protein N-terminus binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0