Search
Search

Tfap2c| Gene Symbol: Tfap2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159696.1 | NP_001153168.1 | 7 | 1363 | ||
| NM_009335.2 | NP_033361.2 | 7 | 1422 | |||
| GenBank mRNA | AK050973.1 | - | 7 | 1340 | ||
| AK141211.1 | - | 7 | 1324 | |||
| AK145166.1 | - | 7 | 1372 | |||
| AK145603.1 | - | 7 | 1317 | |||
| BC003778.1 | - | 7 | 1296 | 3592762 | ||
| KJ463836.1 | - | 7 | 1218 | |||
| X94694.1 | - | 7 | 1401 | |||