Search
Search

TESK2| Gene Symbol: TESK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320800.1 | NP_001307729.1 | 4 | 518 | ||
| NM_007170.2 | NP_009101.2 | 5 | 826 | |||
| XM_006710287.2 | XP_006710350.1 | 6 | 866 | |||
| XM_011540497.1 | XP_011538799.1 | 7 | 899 | |||
| XM_011540500.2 | XP_011538802.1 | 5 | 798 | |||
| XM_017000048.1 | XP_016855537.1 | 6 | 1154 | |||
| XM_017000049.1 | XP_016855538.1 | 4 | 951 | |||
| GenBank mRNA | AB057597.1 | - | 5 | 818 | ||
| AJ132545.1 | - | 4 | 770 | |||
| AK027573.1 | - | 5 | 800 | |||
| AK294172.1 | - | 4 | 493 | |||
| BC022194.1 | - | 5 | 806 | |||
| BC033085.1 | - | 4 | 437 | 5425494 | ||