Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ATG7 SIRNA Assays ›
          Gene Symbol:
          ATG7
          Gene Name:
          autophagy related 7
          Gene Aliases:
          APG7-LIKE, APG7L, GSA7
          Chromosome Location:
          Chr.3: 11272324 - 11564704 on Build GRCh38
          UniGene ID:
          Hs.38032
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s20650

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ATG7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001136031.2 NP_001129503.2 4 378
          NM_001144912.1 NP_001138384.1 4 378
          NM_006395.2 NP_006386.1 4 378
          XM_006712931.3 XP_006712994.1 4 321
          XM_006712932.3 XP_006712995.1 5 585
          XM_006712933.3 XP_006712996.1 4 558
          XM_011533277.2 XP_011531579.1 6 709
          XM_011533278.2 XP_011531580.1 5 414
          XM_011533279.2 XP_011531581.1 6 709
          XM_011533280.2 XP_011531582.1 6 709
          XM_011533281.2 XP_011531583.1 6 709
          XM_011533282.2 XP_011531584.1 6 709
          XM_011533283.2 XP_011531585.1 6 709
          XM_011533284.2 XP_011531586.1 6 709
          XM_011533285.2 XP_011531587.1 6 709
          XM_011533286.2 XP_011531588.1 6 709
          XM_017005542.1 XP_016861031.1 4 295
          XM_017005543.1 XP_016861032.1 4 321
          XM_017005544.1 XP_016861033.1 5 583
          XM_017005545.1 XP_016861034.1 4 363
          XM_017005546.1 XP_016861035.1 5 414
          XM_017005547.1 XP_016861036.1 6 709
          XM_017005548.1 XP_016861037.1 4 321
          XM_017005549.1 XP_016861038.1 5 592
          XM_017005550.1 XP_016861039.1 4 321
          XM_017005551.1 XP_016861040.1 4 321
          XM_017005552.1 XP_016861041.1 4 321
          XM_017005553.1 XP_016861042.1 2 73
          XM_017005554.1 XP_016861043.1 2 73
          GenBank mRNA AF094516.1 - 3 286
          AK075221.1 - 4 305
          AK298833.1 - 4 322
          AK303694.1 - 4 305
          AK314504.1 - 4 295
          BC000091.2 - 4 293 3504204
          DA177126.1 - 4 380

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular protein modification process
          protein lipidation
          C-terminal protein lipidation
          cellular response to nitrogen starvation
          cellular response to starvation
          positive regulation of autophagy
          protein transport
          macroautophagy
          positive regulation of macroautophagy
          protein ubiquitination
          positive regulation of protein modification process
          piecemeal microautophagy of nucleus
          suppression by virus of host autophagy
          positive regulation of apoptotic process
          late nucleophagy
          positive regulation of protein catabolic process
          defense response to virus
          membrane fusion
          cellular response to hyperoxia
          regulation of mitophagy
          ubiquitin activating enzyme activity
          protein binding
          transcription factor binding
          Atg12 activating enzyme activity
          Atg8 activating enzyme activity
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0