Search
Search

Ahcyl2| Gene Symbol: Ahcyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171000.2 | NP_001164471.2 | 7 | 1046 | ||
| NM_001171001.1 | NP_001164472.1 | 7 | 766 | |||
| NM_021414.6 | NP_067389.5 | 7 | 1049 | |||
| GenBank mRNA | AK014539.1 | - | 7 | 997 | ||
| AK053527.1 | - | 7 | 730 | |||
| AK122382.1 | - | 5 | 752 | |||
| BC027436.1 | - | 7 | 961 | |||
| BC059255.1 | - | 3 | 220 | |||
| BC079660.1 | - | 7 | 959 | 6853550 | ||