Search
Search

Ttc30a1| Gene Symbol: Ttc30a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030188.3 | NP_084464.3 | 1 | 973 | ||
| GenBank mRNA | AK019670.1 | - | 1 | 847 | ||
| AK029535.1 | - | 776 | ||||
| AK045096.1 | - | 1 | 773 | |||
| BC004773.1 | - | 1 | 768 | 3256753 | ||