Search
Search

AHSA1| Gene Symbol: AHSA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321441.1 | NP_001308370.1 | 2 | 144 | ||
| NM_012111.2 | NP_036243.1 | 2 | 278 | |||
| GenBank mRNA | AF164791.1 | - | 2 | 184 | ||
| AJ243310.1 | - | 2 | 249 | |||
| AK300766.1 | - | 2 | 276 | |||
| AK313824.1 | - | 2 | 212 | |||
| BC000321.2 | - | 2 | 226 | 2821759 | ||
| BG432594.1 | - | 2 | 279 | |||
| BQ073652.1 | - | 2 | 68 | |||
| CX786861.1 | - | 2 | 155 | |||