Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CELF1 SIRNA Assays ›
Gene Symbol:
CELF1
Gene Name:
CUGBP, Elav-like family member 1
Gene Aliases:
BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50
Chromosome Location:
Chr.11: 47465937 - 47565569 on Build GRCh38
UniGene ID:
Hs.595333
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s20956
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CELF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025596.2
NP_001020767.1
1
88
NM_001172639.1
NP_001166110.1
4
1053
NM_001172640.1
NP_001166111.1
1
88
NM_006560.3
NP_006551.1
2
337
NM_198700.2
NP_941989.1
2
279
XM_011519848.2
XP_011518150.1
2
160
XM_011519850.2
XP_011518152.2
2
399
XM_011519852.2
XP_011518154.1
4
508
XM_011519854.1
XP_011518156.1
6
616
XM_011519855.1
XP_011518157.1
2
337
XM_011519856.1
XP_011518158.1
3
418
XM_011519857.1
XP_011518159.1
2
289
XM_011519858.1
XP_011518160.1
2
384
XM_011519859.1
XP_011518161.1
6
616
XM_017017101.1
XP_016872590.1
5
500
XM_017017102.1
XP_016872591.1
4
419
XM_017017103.1
XP_016872592.1
3
312
XM_017017104.1
XP_016872593.1
3
312
XM_017017105.1
XP_016872594.1
3
312
XM_017017106.1
XP_016872595.1
5
500
XM_017017107.1
XP_016872596.1
3
312
XM_017017108.1
XP_016872597.1
3
312
XM_017017109.1
XP_016872598.1
2
399
XM_017017110.1
XP_016872599.1
3
312
XM_017017111.1
XP_016872600.1
3
312
XM_017017112.1
XP_016872601.1
3
312
XM_017017113.1
XP_016872602.1
3
312
XM_017017114.1
XP_016872603.1
3
312
XM_017017115.1
XP_016872604.1
4
340
XM_017017116.1
XP_016872605.1
3
375
XM_017017117.1
XP_016872606.1
2
136
XM_017017118.1
XP_016872607.1
2
258
XM_017017119.1
XP_016872608.1
4
477
XM_017017120.1
XP_016872609.1
2
289
XM_017017121.1
XP_016872610.1
2
384
XM_017017122.1
XP_016872611.1
2
289
XM_017017123.1
XP_016872612.1
2
337
XM_017017124.1
XP_016872613.1
4
431
XM_017017125.1
XP_016872614.1
2
160
XM_017017126.1
XP_016872615.1
2
289
XM_017017127.1
XP_016872616.1
2
313
XM_017017128.1
XP_016872617.1
2
337
XM_017017129.1
XP_016872618.1
3
335
XM_017017130.1
XP_016872619.1
2
289
XM_017017131.1
XP_016872620.1
2
313
XM_017017132.1
XP_016872621.1
2
337
XM_017017133.1
XP_016872622.1
2
128
XM_017017134.1
XP_016872623.1
3
267
XM_017017135.1
XP_016872624.1
2
289
GenBank mRNA
AB210019.1
-
4
1053
AF248648.1
-
1
96
AF267533.1
-
1
89
AF267534.1
-
1
89
AJ007988.1
-
1
78
AK304430.1
-
1
31
BC031079.1
-
2
279
5297141
CB960956.1
-
3
240
DA217269.1
-
4
360
DA320861.1
-
2
338
DB162286.1
-
2
237
U63289.1
-
2
215
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splice site selection
mRNA processing
germ cell development
spermatid development
embryo development
RNA interference
negative regulation of translation
positive regulation of multicellular organism growth
regulation of RNA splicing
nucleotide binding
translation repressor activity, nucleic acid binding
RNA binding
mRNA binding
protein binding
translation initiation factor binding
BRE binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0