Search
Search

DCTN6| Gene Symbol: DCTN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006571.3 | NP_006562.1 | 3, 4 | 273 | ||
| GenBank mRNA | AK314128.1 | - | 3, 4 | 230 | ||
| BC013175.1 | - | 3, 4 | 217 | 3925231 | ||
| BI562535.1 | - | 3, 4 | 259 | |||
| CR456851.1 | - | 3, 4 | 186 | |||
| D84145.1 | - | 3, 4 | 273 | |||