Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZMYND11 SIRNA Assays ›
Gene Symbol:
ZMYND11
Gene Name:
zinc finger MYND-type containing 11
Gene Aliases:
BRAM1, BS69, MRD30
Chromosome Location:
Chr.10: 134340 - 254637 on Build GRCh38
UniGene ID:
Hs.292265
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s21153
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZMYND11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001202464.1
NP_001189393.1
10
955
NM_001202465.1
NP_001189394.1
9
1195
NM_001202466.1
NP_001189395.1
10
1285
NM_001202467.1
NP_001189396.1
9
879
NM_001202468.1
NP_001189397.1
10
1041
NM_006624.5
NP_006615.2
11
1363
NM_212479.3
NP_997644.2
11
1447
XM_005252359.4
XP_005252416.1
11
1203
XM_005252361.3
XP_005252418.1
10
1124
XM_005252362.2
XP_005252419.1
10
1288
XM_006717376.2
XP_006717439.1
10
1326
XM_017015587.1
XP_016871076.1
11
1116
XM_017015588.1
XP_016871077.1
11
1352
XM_017015589.1
XP_016871078.1
11
1193
XM_017015590.1
XP_016871079.1
11
1650
XM_017015591.1
XP_016871080.1
12
1163
XM_017015592.1
XP_016871081.1
10
1031
XM_017015593.1
XP_016871082.1
10
1471
XM_017015594.1
XP_016871083.1
10
959
GenBank mRNA
AK294469.1
-
9
877
AK294781.1
-
10
967
AK296427.1
-
9
851
AK296518.1
-
6
538
AK312570.1
-
11
1100
AY732489.1
-
7
662
BC012586.2
-
10
1042
BC034784.1
-
11
1228
5171494
BC091489.1
-
10
955
6186814
DQ335452.1
-
10
1022
DQ335453.1
-
9
860
DQ335454.1
-
9
860
DQ335455.1
-
10
1022
X86098.1
-
11
1146
Back To Top
More Information
Panther Classification:
Molecular Function -
transcription cofactor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
cell cycle
cell proliferation
viral process
covalent chromatin modification
regulation of transcription elongation from RNA polymerase II promoter
negative regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of JNK cascade
negative regulation of extrinsic apoptotic signaling pathway
RNA polymerase II regulatory region sequence-specific DNA binding
chromatin binding
transcription cofactor activity
transcription corepressor activity
protein binding
zinc ion binding
methylated histone binding
histone binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0