Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARPP21 SIRNA Assays ›
Gene Symbol:
ARPP21
Gene Name:
cAMP regulated phosphoprotein 21
Gene Aliases:
ARPP-21, R3HDM3, RCS, TARPP
Chromosome Location:
Chr.3: 35638981 - 35798933 on Build GRCh38
UniGene ID:
Hs.475902
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s21170
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARPP21
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025068.1
NP_001020239.1
3
359
NM_001025069.1
NP_001020240.1
3
368
NM_001267616.1
NP_001254545.1
3
1515
NM_001267617.1
NP_001254546.1
3
325
NM_001267618.1
NP_001254547.1
2
456
NM_001267619.1
NP_001254548.1
2
290
NM_016300.4
NP_057384.2
3
342
NM_198399.1
NP_938409.1
2
350
XM_005264811.2
XP_005264868.1
3
371
XM_006712943.2
XP_006713006.1
3
371
XM_006712944.2
XP_006713007.1
3
371
XM_011533298.2
XP_011531600.1
3
371
XM_011533299.2
XP_011531601.1
3
915
XM_011533300.2
XP_011531602.1
3
534
XM_011533301.2
XP_011531603.1
3
365
XM_011533302.2
XP_011531604.1
4
457
XM_011533303.2
XP_011531605.1
4
1007
XM_017005574.1
XP_016861063.1
3
461
XM_017005575.1
XP_016861064.1
4
12094
XM_017005576.1
XP_016861065.1
3
915
XM_017005577.1
XP_016861066.1
3
365
XM_017005578.1
XP_016861067.1
3
341
XM_017005579.1
XP_016861068.1
3
915
XM_017005580.1
XP_016861069.1
4
1007
XM_017005581.1
XP_016861070.1
4
12094
XM_017005582.1
XP_016861071.1
3
915
XM_017005583.1
XP_016861072.1
3
365
XM_017005584.1
XP_016861073.1
3
915
XM_017005585.1
XP_016861074.1
3
915
XM_017005586.1
XP_016861075.1
3
371
XM_017005587.1
XP_016861076.1
3
534
XM_017005588.1
XP_016861077.1
4
1007
XM_017005589.1
XP_016861078.1
4
457
XM_017005590.1
XP_016861079.1
4
12094
XM_017005591.1
XP_016861080.1
3
915
XM_017005592.1
XP_016861081.1
3
915
XM_017005593.1
XP_016861082.1
3
461
XM_017005594.1
XP_016861083.1
3
365
XM_017005595.1
XP_016861084.1
4
1007
XM_017005596.1
XP_016861085.1
3
915
XM_017005597.1
XP_016861086.1
3
915
XM_017005598.1
XP_016861087.1
4
1007
XM_017005599.1
XP_016861088.1
3
915
XM_017005600.1
XP_016861089.1
3
534
XM_017005601.1
XP_016861090.1
3
365
XM_017005602.1
XP_016861091.1
3
915
XM_017005603.1
XP_016861092.1
3
534
XM_017005604.1
XP_016861093.1
3
365
XM_017005605.1
XP_016861094.1
4
1007
XM_017005606.1
XP_016861095.1
4
457
XM_017005607.1
XP_016861096.1
3
915
XM_017005608.1
XP_016861097.1
3
915
XM_017005609.1
XP_016861098.1
4
1007
XM_017005610.1
XP_016861099.1
3
915
XM_017005611.1
XP_016861100.1
3
915
XM_017005612.1
XP_016861101.1
3
915
GenBank mRNA
AF112220.1
-
2
315
AK055064.1
-
3
368
AK289649.1
-
2
305
AK289868.1
-
3
362
AK294484.1
-
2
303
AV718064.1
-
3
359
AV718166.1
-
3
368
BC017805.1
-
2
350
BC036399.1
-
3
330
5263894
BC041385.1
-
3
1103
BC051828.1
-
3
342
6012704
BC066651.1
-
1
869
BG535216.1
-
2
351
BX488342.1
-
2
454
DA275579.1
-
2
456
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
cellular response to heat
molecular_function
nucleic acid binding
calmodulin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0