Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BTK SIRNA Assays ›
          Gene Symbol:
          BTK
          Gene Name:
          Bruton tyrosine kinase
          Gene Aliases:
          AGMX1, AT, ATK, BPK, IMD1, PSCTK1, XLA
          Chromosome Location:
          Chr.X: 101349447 - 101390796 on Build GRCh38
          UniGene ID:
          Hs.159494
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s2124

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BTK
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000061.2 NP_000052.1 16 1775
          NM_001287344.1 NP_001274273.1 16 1931
          NM_001287345.1 NP_001274274.1 14 1412
          GenBank mRNA AF153364.1 - 2 127
          AF153756.1 - 2 127
          AF153757.1 - 2 127
          AF153758.1 - 2 127
          AF153759.1 - 2 127
          AF153760.1 - 2 127
          AF153761.1 - 2 127
          AF153762.1 - 2 127
          AF153764.1 - 2 123
          AJ888377.1 - 4 336
          AJ888378.1 - 4 382
          AJ888381.1 - 3 220
          AK057105.1 - 14 1366
          AK309851.1 - 16 1917
          AK314382.1 - 16 1742
          AM051275.1 - 4 337
          AM051276.1 - 5 595
          AM051280.1 - 3 229
          BC109079.1 - 16 1707 40034287
          BC109080.1 - 16 1707 40034289
          KF241986.1 - 15 1582
          X58957.1 - 16 1714

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_013482, L08967, L10627, L29788, BC053392.

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of cytokine production
          adaptive immune response
          regulation of B cell cytokine production
          MyD88-dependent toll-like receptor signaling pathway
          regulation of B cell apoptotic process
          transcription, DNA-templated
          protein phosphorylation
          transmembrane receptor protein tyrosine kinase signaling pathway
          I-kappaB kinase/NF-kappaB signaling
          mesoderm development
          peptidyl-tyrosine phosphorylation
          calcium-mediated signaling
          intracellular signal transduction
          peptidyl-tyrosine autophosphorylation
          Fc-epsilon receptor signaling pathway
          B cell activation
          regulation of cell proliferation
          innate immune response
          positive regulation of B cell differentiation
          cell maturation
          B cell receptor signaling pathway
          positive regulation of NF-kappaB transcription factor activity
          apoptotic signaling pathway
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          receptor binding
          protein binding
          ATP binding
          phosphatidylinositol-3,4,5-trisphosphate binding
          identical protein binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0