Search
Search

SUPT16H| Gene Symbol: SUPT16H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007192.3 | NP_009123.1 | 26 | 3437 | ||
| XM_011536381.2 | XP_011534683.1 | 27 | 3439 | |||
| GenBank mRNA | AF164924.1 | - | 6 | 698 | ||
| AK001719.1 | - | 26 | 3173 | |||
| AK091676.1 | - | 6 | 1084 | |||