Search
Search

Rbpj| Gene Symbol: Rbpj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106631.1 | NP_001100101.1 | 10 | 988 | ||
| XM_008770190.1 | XP_008768412.1 | 9 | 1659 | |||
| XM_008770191.1 | XP_008768413.1 | 9 | 900 | |||
| XM_008770192.1 | XP_008768414.1 | 9 | 3078 | |||
| XM_008770193.1 | XP_008768415.1 | 10 | 1071 | |||